All Photos(2)
About This Item
Linear Formula:
BrC6H3F2
CAS Number:
Molecular Weight:
192.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
refractive index
n20/D 1.499 (lit.)
bp
140 °C (lit.)
density
1.676 g/mL at 25 °C (lit.)
functional group
bromo
fluoro
SMILES string
Fc1cc(F)cc(Br)c1
InChI
1S/C6H3BrF2/c7-4-1-5(8)3-6(9)2-4/h1-3H
InChI key
JHLKSIOJYMGSMB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Bromo-3,5-difluorobenzene was used in the synthesis of 4-bromo-2,6-difluorobenzoic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
111.2 °F - closed cup
Flash Point(C)
44 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Regioselective ortho-lithiation of chloro and bromo substituted fluoroarenes.
Tetrahedron Letters, 37(36), 6551- 6554 (1996)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service