Skip to Content
Merck
All Photos(1)

Key Documents

28700

Sigma-Aldrich

Cyclododecanecarboxylic acid

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O2
CAS Number:
Molecular Weight:
212.33
Beilstein:
1869970
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

solid

mp

97-98 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1CCCCCCCCCCC1

InChI

1S/C13H24O2/c14-13(15)12-10-8-6-4-2-1-3-5-7-9-11-12/h12H,1-11H2,(H,14,15)

InChI key

JWIPDQOXAJMVHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Cyclododecanecarboxylic acid may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service