Skip to Content
Merck
All Photos(1)

Documents

283754

Sigma-Aldrich

2(5H)-Furanone

98%

Synonym(s):

γ-Crotonolactone, 2-Buten-1,4-olide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O2
CAS Number:
Molecular Weight:
84.07
Beilstein:
383585
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.469 (lit.)

bp

86-87 °C/12 mmHg (lit.)

mp

4-5 °C (lit.)

density

1.185 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1OCC=C1

InChI

1S/C4H4O2/c5-4-2-1-3-6-4/h1-2H,3H2

InChI key

VIHAEDVKXSOUAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chiral urea compounds catalyzed hetero-Michael addition reaction of 2(5H)-furanone (γ-crotonolactone) to pyrrolidine. The quorum sensing inhibition activity by 2(5H)-furanone was studied using bioindicator strains.

Application

2(5H)-Furanone (γ-Crotonolactone) has been used in:
  • synthesis of (+)-L-733,060, (+)-CP-99,994 and (2S,3R)-3-hydroxypipecolic acid
  • synthesis of 5-substituted 2(5H) furanones (γ-butenolides) via direct aldol reaction with aromatic aldehydes catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts
  • Michael addition reactions for synthesis of lignans
  • three-component Michael-Aldol reactions with an aldehyde anda thiolate or carbanion

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 49, 9039-9039 (1993)
Tetrahedron, 49, 8073-8073 (1993)
Yoshihiro Sohtome et al.
Chemical & pharmaceutical bulletin, 52(4), 477-480 (2004-04-02)
Chiral urea compounds 10a-g were synthesized as catalysts for conjugate addition of pyrrolidine (2) to gamma-crotonolactone (3). In the presence of a catalytic amount of the chiral ureas, this hetero-Michael reaction was greatly accelerated. Asymmetric induction was observed with the
Tetrahedron, 49, 4173-4173 (1993)
Rebecca J Chesterfield et al.
ACS synthetic biology, 9(8), 2107-2118 (2020-08-14)
Strigolactones are plant hormones and rhizosphere signaling molecules with key roles in plant development, mycorrhizal fungal symbioses, and plant parasitism. Currently, sensitive, specific, and high-throughput methods of detecting strigolactones are limited. Here, we developed genetically encoded fluorescent strigolactone biosensors based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service