Skip to Content
Merck
All Photos(1)

Documents

282286

Sigma-Aldrich

1,4-Diiodotetrafluorobenzene

98%

Synonym(s):

1,2,4,5-Tetrafluoro-3,6-diiodobenzene, 1,4-Diiodo-2,3,5,6-tetrafluorobenzene, 3,6-Diiodo-1,2,4,5-tetrafluorobenzene, Tetrafluoro-1,4-diiodobenzene, p-Diiodoperfluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6F4I2
CAS Number:
Molecular Weight:
401.87
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

108-110 °C (lit.)

SMILES string

Fc1c(F)c(I)c(F)c(F)c1I

InChI

1S/C6F4I2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

VIXRAZODEODOJF-UHFFFAOYSA-N

General description

1,4-Diiodotetrafluorobenzene forms trimeric complexes as new, halogen-bonded mesogens with two molecules of alkoxystilbazole. The electron density study of the halogen-bonded complex of 4,4′-dipyridyl-N,N′-dioxide with 1,4-diiodotetrafluorobenzene at 90K has been reported.

1,4-Diiodotetrafluorobenzene is a ditopic perfluorinated iodobenzene used for cocrystallization of halopyridinium salts.

Application

1,4-Diiodotetrafluorobenzene has been used as the halogen bonding donor during the preparation of phosphorescent cocrystal with polycyclic aromatic hydrocarbons.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halogen and hydrogen bond motifs in ionic cocrystals derived from 3-halopyridinium halogenides and perfluorinated iodobenzenes
Lidija P et al.
Crystal Growth & Design, 21, 6044-6050 (2021)
Jay Quentin et al.
Molecules (Basel, Switzerland), 25(4) (2020-02-23)
The halogen-bond (X-bond) donors 1,3- and 1,4-diiodotetrafluorobenzene (1,3-di-I-tFb and 1,4-di-I-tFb, respectively) form cocrystals with trans-1,2-bis(2-pyridyl)ethylene (2,2'-bpe) assembled by N···I X-bonds. In each cocrystal, 2(1,3-di-I-tFb)·2(2,2'-bpe) and (1,4-di-I-tFb)·(2,2'-bpe), the donor molecules support the C=C bonds of 2,2'-bpe to undergo an intermolecular [2+2]
Mesogenic, trimeric, halogen-bonded complexes from alkoxystilbazoles and 1, 4-diiodotetrafluorobenzene.
<BIG>Bruce DW, et al. </BIG>
New. J. Chem., 32(3), 477-482 (2008)
Phosphorescent co-crystal assembled by 1, 4-diiodotetrafluorobenzene with carbazole based on C-Ip halogen bonding.
<BIG>Gao HY, et al. </BIG>
Journal of Materials Chemistry, 22(12), 5336-5343 (2012)
Jiehao Fu et al.
iScience, 23(3), 100965-100965 (2020-03-22)
Here we introduce a σ-hole-containing volatile solid additive, 1, 4-diiodotetrafluorobenzene (A3), in PM6:Y6-based OSCs. Aside from the appropriate volatility of A3 additive, the synergetic halogen interactions between A3 and photoactive matrix contribute to more condensed and ordered molecular arrangement in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service