281646
2,3-Bis(2-pyridyl)pyrazine
98%
Synonym(s):
2,3-Di-2-pyridylpyrazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H10N4
CAS Number:
Molecular Weight:
234.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
168-170 °C (lit.)
SMILES string
c1ccc(nc1)-c2nccnc2-c3ccccn3
InChI
1S/C14H10N4/c1-3-7-15-11(5-1)13-14(18-10-9-17-13)12-6-2-4-8-16-12/h1-10H
InChI key
WTZDTUCKEBDJIM-UHFFFAOYSA-N
Related Categories
Application
2,3-Bis(2-pyridyl)pyrazine (dpp) has been employed:
- as building block for supermolecular molecules
- in the preparation of luminescent complexes Ru(bpy)2(dpp)2+ and (bpy)2Ru-(dpp)Ru(bpp)24+ [bpy=2,2′-bipyridine]
- in the preparation of binary and ternary systems with V(IV)O and Cu(II) ions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Inorganic Chemistry, 32, 3003-3003 (1993)
Spectroscopic and electrochemical properties of the dimer tetrakis (2, 2'-bipyridine)(. mu.-2, 3-bis (2-pyridyl) pyrazine) diruthenium (II) and its monomeric analog.
Braunstein CH, et al.
Inorganic Chemistry, 23(7), 857-864 (1984)
Jing Xiang et al.
Dalton transactions (Cambridge, England : 2003), 48(2), 741-750 (2018-12-19)
A series of polynuclear metal complexes, [Cu2(L1)(PPh3)4](ClO4)2 (1), [Cu3(L2)(PPh3)6](ClO4) (2), [Cu3(L3)(PPh3)6] (3), [Ag2(L1)(PPh3)4](BF4)2 (4), [Ag4(L2)2(PPh3)6] (5) and [Ag3(L3)(PPh3)5] (6), have been obtained from the reactions of the highly conjugated bridging ligands 2,3-bis(2-pyridyl)pyrazine (L1), 2,3-bis(2-tetrazoyl)pyrazine (H2L2) and 2,3-bis(2-tetrazoyl)imidazole (H3L3) with [Cu(MeCN)4]ClO4
Daniele Sanna et al.
Dalton transactions (Cambridge, England : 2003), 41(41), 12824-12838 (2012-09-15)
The binary and ternary systems formed by V(IV)O and Cu(II) ions with ligands (L) based on the pyridine ring (1,10-phenanthroline (phen), 2,2'-bipyridine (bpy), 2,2':6',2''-terpyridine (terpy), 2,2'-bipyrimidine (bpm) and 2,3-bis(2-pyridyl)pyrazine (bpp)) were studied, combining spectroscopic (EPR and UV-vis), pH-potentiometric and computational
Cléia Justino Nunes et al.
Journal of inorganic biochemistry, 149, 49-58 (2015-05-30)
In this work, the influence of two new dinuclear copper(II) complexes in the viability of melanoma cells (B16F10 and TM1MNG3) was investigated, with the aim of verifying possible correlations between their cytotoxicity and their structure. One of the complexes had
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service