Skip to Content
Merck
All Photos(1)

Key Documents

280348

Sigma-Aldrich

2-Bromooctanoic acid

97%

Synonym(s):

2-Bromocaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(Br)COOH
CAS Number:
Molecular Weight:
223.11
Beilstein:
1760958
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

contains

Silver Wool as stabilizer

refractive index

n20/D 1.471 (lit.)

bp

140 °C/5 mmHg (lit.)

density

1.278 g/mL at 25 °C (lit.)

functional group

bromo
carboxylic acid

SMILES string

CCCCCCC(Br)C(O)=O

InChI

1S/C8H15BrO2/c1-2-3-4-5-6-7(9)8(10)11/h7H,2-6H2,1H3,(H,10,11)

InChI key

GTGTXZRPJHDASG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromooctanoic acid is an effective substitute for the expensive (2,000 times) and cell-growth-inhibiting polyhydroxyalkanoic acid synthesis inhibitor, cerulenin. It blocks the formation of polyhydroxyalkanoic acid in Pseudomonas fluorescens BM07 without any influence on the cell growth when grown on fructose. It inhibits β-oxidation of fatty acids in perfused rat liver and in mitochondria isolated from rat liver.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Channeling of intermediates derived from medium-chain fatty acids and de novo-synthesized fatty acids to polyhydroxyalkanoic acid by 2-bromooctanoic acid in Pseudomonas fluorescens BM07.
LEE H-J, et al.
Journal of microbiology and biotechnology, 14(6), 1256-1266 (2004)
B M Raaka et al.
The Journal of biological chemistry, 254(14), 6755-6762 (1979-07-25)
Incubation of rat liver mitochondria with 10 microM DL-2-bromooctanoate causes complete and irreversible inactivation of 3-ketothiolase I (acyl-CoA:acetyl-CoA C-acyltransferase). Evidence is presented that mitochondria convert bromooctanoate to 2-bromo-3-ketooctanoyl-CoA, an alpha-haloketone which is probably the active form of the inhibitor. The
N Mayorek et al.
European journal of biochemistry, 182(2), 395-400 (1989-06-15)
The limiting role of diacylglycerol acyltransferase with respect to triacylglycerol synthesis in cultured rat hepatocytes was evaluated by following the inhibition of the overall synthetic flux by 2-bromooctanoate acting as an inhibitor of the diacylglycerol acyltransferase step. The flux-control coefficient
Karen M Tobin et al.
FEMS microbiology letters, 253(1), 111-118 (2005-11-02)
A number of Pseudomonas strains accumulated polyhdroxyalkanoate (PHA) from a variety of aromatic hydrocarbons. In many strains the level of PHA accumulation was dependent on the side chain length of the phenylalkanoic acid provided for growth. 4 of the 8
J G Conway et al.
Cancer research, 47(18), 4795-4800 (1987-09-15)
The diffusion of H2O2 into the cytoplasm from peroxisomes during high rates of peroxisomal beta oxidation of fatty acids was studied in perfused livers from rats treated with the hepatocarcinogenic peroxisome proliferator, nafenopin. Efflux of oxidized glutathione (GSSG) into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service