Skip to Content
Merck
All Photos(1)

Key Documents

278610

Sigma-Aldrich

Moroxydine hydrochloride

99%

Synonym(s):

N-(Aminoiminomethyl)-4-morpholinecarboximidamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13N5O · HCl
CAS Number:
Molecular Weight:
207.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

211-214 °C (lit.)

functional group

amine
ether

SMILES string

Cl[H].NC(=N)NC(=N)N1CCOCC1

InChI

1S/C6H13N5O.ClH/c7-5(8)10-6(9)11-1-3-12-4-2-11;/h1-4H2,(H5,7,8,9,10);1H

InChI key

FXYZDFSNBBOHTA-UHFFFAOYSA-N

General description

Determination of moroxydine hydrochloride in Gan Mao Qing capsules by derivative spectrophotometric signal multiplier method has been reported. Therapeutical efficiency of moroxidine hydrochloride against herpes virus induced keratoconjunctivitis has been evaluated by immunofluorescence techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Recurrent herpetic gingivostomatitis treated with moroxydine, antiherpin and the immunostimulant SRE].
V Topciu et al.
Bacteriologia, virusologia, parazitologia, epidemiologia (Bucharest, Romania : 1990), 37(3-4), 65-67 (1992-07-01)
P Athanasiu et al.
Revue roumaine de virologie (Bucharest, Romania : 1990), 44(1-2), 17-20 (1993-01-01)
Treatment with moroxidine, Romanian preparation with virustatic effects, was applied in 350 patients with different localisation herpes zoster lesions. Treatment had good effects, especially when it was applied early.
V Topciu et al.
Revue roumaine de virologie (Bucharest, Romania : 1990), 44(1-2), 91-95 (1993-01-01)
A group of 16 patients with ophthalmic zona zoster received antiviral and immunostimulating treatment with Romanian specific products. Results were spectacular. The problem of the identity of the varicella and zoster viruses is discussed.
S Sheppard
Acta dermato-venereologica. Supplementum, 183, 1-9 (1994-01-01)
A review is presented which began as a literature survey listing verbatim over fifty abstracts and references to the use of moroxydine in the treatment of a number of viral disorders throughout Europe and Scandinavia between 1960 and 1985. The
[Virostatic effect in the treatment of pityriasis rosea irritata with moroxydine-HCI].
J W Grobe
Dermatosen in Beruf und Umwelt. Occupation and environment, 32(1), 25-25 (1984-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service