Skip to Content
Merck
All Photos(3)

Key Documents

27370

Sigma-Aldrich

Cinchonine

crystallized, ≥98.0% (NT)

Synonym(s):

NSC 6176

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
Beilstein:
89689
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

powder

optical activity

[α]20/D +225±5°, c = 0.5% in ethanol

quality

crystallized

impurities

~15% dihydrocinchonine (HPLC)

mp

260-263 °C

functional group

hydroxyl

SMILES string

O[C@H]([C@H]1C[C@@H]2CCN1C[C@@H]2C=C)c3ccnc4ccccc34

InChI

1S/C19H22N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h2-7,9,13-14,18-19,22H,1,8,10-12H2/t13-,14-,18+,19-/m0/s1

InChI key

KMPWYEUPVWOPIM-QAMTZSDWSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

General description

Cinchonine is a cinchona alkaloid generally found in the bark of Cinchona officinalis plants. It is a pseudoenantiomer that is commonly employed in malaria therapy. Cinchonine is also used as an organocatalyst in many asymmetric reactions.

Application

Cinchonine can be used:
  • As a substrate in the study of supercritical fluid chromatography (SFC) parameters in pharmaceutical analysis.
  • To prepare possible butyrylcholinesterase inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Chemistry and biology of Cinchona alkaloids
Kacprzak K
Natural Products, 605-641 (2013)
Anita Bosak et al.
PloS one, 13(10), e0205193-e0205193 (2018-10-06)
This paper describes the synthesis and anticholinesterase potency of Cinchona-based alkaloids; ten quaternary derivatives of cinchonines and their corresponding pseudo-enantiomeric cinchonidines. The quaternization of quinuclidine moiety of each compound was carried out with groups diverse in their size: methyl, benzyl
Vibrational analysis of cinchona alkaloids in the solid state and aqueous solutions
Roman M, et al.
Journal of Raman Spectroscopy, 46(11), 1041-1052 (2015)
Gold nanoparticles decorated with a cinchonine organocatalyst: application in the asymmetric alpha-amination of beta-ketoesters
Santacruz L, et al.
New. J. Chem., 38(2), 636-640 (2014)
Chensong Pan et al.
Journal of the American Society for Mass Spectrometry, 16(2), 263-270 (2005-02-08)
A method with carbon nanotubes functioning both as the adsorbent of solid-phase extraction (SPE) and the matrix for matrix assisted laser desorption/ionization mass spectrometry (MALDI-MS) to analyze small molecules in solution has been developed. In this method, 10 microL suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service