Skip to Content
Merck
All Photos(1)

Documents

262528

Sigma-Aldrich

N,N-Diisopropylmethylphosphonamidic chloride

95%

Synonym(s):

Chloro(diisopropylamino)methoxyphosphine, Methyl N,N-diisopropylchlorophosphoramidite, Methyl N,N-diisopropylphosphoramidochloridite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NP(OCH3)Cl
CAS Number:
Molecular Weight:
197.64
Beilstein:
3600769
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

refractive index

n20/D 1.468 (lit.)

bp

34-35 °C/0.8 mmHg (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

COP(Cl)N(C(C)C)C(C)C

InChI

1S/C7H17ClNOP/c1-6(2)9(7(3)4)11(8)10-5/h6-7H,1-5H3

InChI key

VOVWQRQDSCYAEA-UHFFFAOYSA-N

Application

Reagent used for polymer-supported deoxyoligonucleotide and oligoribonucleotide syntheses.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 747-747 (1993)
Aldrichimica Acta, 16, 47-47 (1983)
Tetrahedron Letters, 34, 2195-2195 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service