Skip to Content
Merck
All Photos(1)

Key Documents

261076

Sigma-Aldrich

Dysprosium

ingot, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Dy
CAS Number:
Molecular Weight:
162.50
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

ingot

reaction suitability

reagent type: catalyst
core: dysprosium

resistivity

89 μΩ-cm, 20°C

bp

2567 °C (lit.)

mp

1412 °C (lit.)

density

8.559 g/mL at 25 °C (lit.)

SMILES string

[Dy]

InChI

1S/Dy

InChI key

KBQHZAAAGSGFKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Li Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(46), 14632-14637 (2012-10-16)
Homochiral Dy(III) complexes: by changing the ligand-to-metal ratio, enantiomeric pairs of a Dy(III) complex of different nuclearity could be obtained. The mono- and dinuclear complexes exhibit characteristics of single-molecule magnets and different slow magnetic relaxation processes. In addition, the dinuclear
Thomas Just Sørensen et al.
Chemical communications (Cambridge, England), 49(8), 783-785 (2012-12-14)
A heterotrimetallic tetranuclear lanthanide complex containing two dysprosium ions, a terbium ion and a europium ion has been prepared by coupling three kinetically stable complexes together using the Ugi reaction. The covalently linked trimetallic system exhibits luminescence from all the
C Manjunatha et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 140-148 (2012-04-07)
CdSiO(3):Dy(3+) (1-9mol%) nanophosphors were prepared for the first time using the solution combustion method. The process of monoclinic phase formation was investigated by PXRD, TG-DTA and FTIR. The results show that the phase formation temperature of combustion-derived monoclinic CdSiO(3) is
Dimitris I Alexandropoulos et al.
Inorganic chemistry, 52(3), 1179-1181 (2013-01-18)
The first use of N-salicylidene-o-aminophenol in 3d/4f chemistry has led to Mn(III)(4)Dy(III)(5) and Mn(III)(4)Dy(III)(3) clusters with unprecedented metal topologies and stoichiometries; both compounds exhibit out-of-phase signals indicative of the slow magnetization relaxation of a single-molecule magnet.
Hai-Hong Wu et al.
Dalton transactions (Cambridge, England : 2003), 42(2), 342-346 (2012-11-15)
The heterometallic appended {MMn(III)(4)} (M = Dy(3+) and K(+)) cubanes were firstly trapped by two diamagnetic POM shells, which were robust enough to construct inorganic crystalline tubular materials. Magnetic study reveals the presence of a SMM-like slow magnetic relaxation feature

Articles

Today, near-room-temperature refrigeration is almost entirely based on a vapor-compression refrigeration cycle.

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service