259071
5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride
≥94% (HPLC)
Synonym(s):
meso-Tetraphenylporphyrin iron(III) chloride
About This Item
Recommended Products
Assay
≥94% (HPLC)
λmax
418 nm
SMILES string
Cl[Fe]1n2c3ccc2c(-c4ccccc4)c5ccc(n5)c(-c6ccccc6)c7ccc(c(-c8ccccc8)c9ccc(n9)c3-c%10ccccc%10)n17
InChI
1S/C44H28N4.ClH.Fe/c1-5-13-29(14-6-1)41-33-21-23-35(45-33)42(30-15-7-2-8-16-30)37-25-27-39(47-37)44(32-19-11-4-12-20-32)40-28-26-38(48-40)43(31-17-9-3-10-18-31)36-24-22-34(41)46-36;;/h1-28H;1H;/q-2;;+3/p-1/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-;;
InChI key
ZDYSAMCSFRQDMN-YKKPBKTHSA-M
Related Categories
Application
- Carbon Nanopowder-Based Stochastic Sensor: This research describes the application of iron(III) porphyrin in carbon nanopowder-based sensors for the ultrasensitive assay of biomarkers in whole blood (Stefan-van Staden et al., 2022).
- High sensitivity carbon monoxide detector: Discusses the development of a carbon monoxide detector using iron tetraphenyl porphyrin functionalized reduced graphene oxide (Shirsat et al., 2023).
- Sulfur-Doped Graphene-Based Electrochemical Sensors: This article examines the use of iron tetraphenyl porphyrin in sensors for detecting limonene in beverages (Niculae et al., 2022).
Features and Benefits
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service