Skip to Content
Merck
All Photos(2)

Documents

254584

Sigma-Aldrich

Crotyl chloride, predominantly trans

95%

Synonym(s):

1-Chloro-2-butene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH2Cl
CAS Number:
Molecular Weight:
90.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

impurities

4% 3-chloro-1-butene

refractive index

n20/D 1.436 (lit.)

bp

84-85 °C (lit.)

density

0.929 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C(C)=C(\[H])CCl

InChI

1S/C4H7Cl/c1-2-3-4-5/h2-3H,4H2,1H3/b3-2+

InChI key

YTKRILODNOEEPX-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Crotyl chloride reacts with trichlorosilane in the presence of tertiary amine and copper salt to give the corresponding allyltrichlorosilane.

Application

Crotyl chloride was used in kinetics measurements of reactions of atomic chlorine with series of unsaturated aldehydes and ketones at 298K. (E)Crotyl chloride was also used in the preparation of (E)-crotyltrichlorosilane.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

23.0 °F

Flash Point(C)

-5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The condensation reaction of trichlorosilane with allylic chlorides catalyzed by copper salts in the presence of a tertiary amine.
Furuya N and Sukawa T.
Journal of Organometallic Chemistry, 96(1), C1-C3 (1975)
Stereohomogeneous synthesis of (E)-and (Z)-crotyltrifluorosilanes and highly stereoselective allylation of aldehydes.
Kira M, et al.
Tetrahedron Letters, 30(9), 1099-1102 (1989)
Rate constants for the reactions of chlorine atoms with a series of unsaturated aldehydes and ketones at 298 K: structure and reactivity.
Wang W, et al.
Physical Chemistry Chemical Physics, 4(10), 1824-1831 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service