Skip to Content
Merck
All Photos(3)

Key Documents

254169

Sigma-Aldrich

Gold(III) chloride hydrate

99.995% trace metals basis

Synonym(s):

Auric chloride, Chloroauric acid, Hydrogen tetrachloroaurate(III) hydrate, Tetrachloroauric(III) acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HAuCl4 · aq
CAS Number:
Molecular Weight:
339.79 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.995% trace metals basis
form:
crystals and lumps

Quality Level

Assay

99.995% trace metals basis

form

crystals and lumps

composition

Degree of hydration, ~3

reaction suitability

reagent type: catalyst
core: gold

technique(s)

ELISA: suitable

impurities

≤55.0 ppm Trace Metal Analysis

density

3.9 g/mL at 25 °C

SMILES string

O.Cl.Cl[Au](Cl)Cl

InChI

1S/Au.4ClH.H2O/h;4*1H;1H2/q+3;;;;;/p-3

InChI key

OVJUDSKPNIBLOO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gold (III) chloride hydrate (HAuCl4) can be used as a precursor to synthesize goldnanoparticles and p-type dopant for most conjugated organic molecules. Thedoping leads to the expansion of crystalline lattice and enhances conductivitydue to the formation of a large number of chargecarriers.

Application

Gold(III) chloride hydrate can be used to prepare TiO2-supported Au-based photocatalyst by deposition-precipitation with urea. These catalysts show a higher amount and production rate of hydrogen via a water-splitting reaction.

Conjugate polythiophene films can be prepared by the solution-doping method using HAuCl4 as a dopant. These highly conductive polymer films can be applied in the field of vapor sensing and organic electronics.

HAuCl4 can be reduced to prepare gold nanoparticles by using different reducing agents like NaBH4, glucose, and poly (propylene imine) dendrimer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo Chuan Huang et al.
Scientific reports, 9(1), 7589-7589 (2019-05-22)
Shewanella oneidensis MR-1, a bioelectricity generating bacterium, is broadly used in bioremediation, microbial fuel cell and dissimilatory reduction and recovery of precious metals. Herein, we report for the first time that photo induction as a trigger to stimulate gold nanoparticles
Fink, J. et al.
Chemistry of Materials, 10, 922-922 (1998)
Ligand-induced gold nanocrystal superlattice formation in colloidal solution.
Lin XM, et al.
Chemistry of Materials, 11(2), 198-202 (1999)
Arvind Pandey et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-03)
It has been suggested that particle size plays an important role in determining the genotoxicity of gold nanoparticles (GNPs). The purpose of this study was to compare the potential radio-sensitization effects of two different sized GNPs (3.9 and 37.4 nm)
The crystal structure of gold(III) chloride.
Clark ES, et al.
Acta Crystallographica, 11, 284-288 (1958)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service