252719
Dodecanophenone
98%
Synonym(s):
1-Phenyl-1-dodecanone
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H5CO(CH2)10CH3
CAS Number:
Molecular Weight:
260.41
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
bp
214-215 °C/16 mmHg (lit.)
mp
45-47 °C (lit.)
SMILES string
CCCCCCCCCCCC(=O)c1ccccc1
InChI
1S/C18H28O/c1-2-3-4-5-6-7-8-9-13-16-18(19)17-14-11-10-12-15-17/h10-12,14-15H,2-9,13,16H2,1H3
InChI key
DJNJZIFFCJTUDS-UHFFFAOYSA-N
Application
Dodecanophenone has been employed as micellar marker in:
- micellar electrokinetic chromatography (MEC) systems
- determination of octanol-water partition coefficients of pharmaceuticals by using pressure-assisted MEC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Megan C Sprinkle et al.
Journal of feline medicine and surgery, 20(2), 83-90 (2017-11-28)
Objectives The aim of this report is to describe the identification of a novel vitamin D metabolite, a C-3, alpha-epimer of 25-hydroxycholecalciferol (3-epi-25(OH)D
Zhongjiang Jia et al.
Journal of chromatography. A, 1007(1-2), 203-208 (2003-08-20)
A rapid screening assay for the determination of octanol-water partition coefficients (log P(OW)) of pharmaceuticals was developed by using pressure-assisted microemulsion electrokinetic chromatography (MEEKC). The microemulsion system contains 50 mM sodium dodecyl sulfate, 0.87 M l-butanol, 82 mM heptane, and
Elisabet Fuguet et al.
Electrophoresis, 23(1), 56-66 (2002-02-05)
A wide study of the compounds and procedures mostly used to determine the electroosmotic flow (EOF) and micelle elution times has been done in seven different micellar electrokinetic chromatography (MEKC) systems. These systems are formed from mixtures of an aqueous
Alejandro Fernández-Pumarega et al.
Analytica chimica acta, 1078, 221-230 (2019-07-31)
Determination of the retention factor of ionized compounds in microemulsion electrokinetic chromatography requires two mobility measurements at the same pH: one in the presence of the microemulsion and another in plain buffer. However, it has been observed that in some
Ali Modir-Rousta et al.
Electrophoresis, 36(7-8), 1016-1023 (2015-02-03)
We recently introduced a pressure-assisted sweeping-reversed migration-EKC (RM-EKC) method for preconcentration of neutral polar N-nitrosamines with low affinity for the micellar phase. The type of surfactant and phase ratio are dominant factors in dictating the magnitude of interactions between analyte
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service