Skip to Content
Merck
All Photos(1)

Key Documents

242527

Sigma-Aldrich

1-Undecene

97%

Synonym(s):

α-Undecene, n-1-Undecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH=CH2
CAS Number:
Molecular Weight:
154.29
Beilstein:
1740044
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.426 (lit.)

bp

192-193 °C (lit.)

mp

−49 °C (lit.)

density

0.75 g/mL at 25 °C (lit.)

functional group

allyl

SMILES string

CCCCCCCCCC=C

InChI

1S/C11H22/c1-3-5-7-9-11-10-8-6-4-2/h3H,1,4-11H2,2H3

InChI key

DCTOHCCUXLBQMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-undecene (or α-undecene) is an organic compound, which belongs to the class of unsaturated aliphatic hydrocarbons. It is a hydrophobic molecule found in the essential oil of Farfugium japonicum and some mushroom species.

Application

1-undecene is an alkene that can be used as a reactant:
  • In the Pd-catalyzed aerobic oxidative amination reaction with nitrogen nucleophiles to produce useful amine analogs.
  • To prepare a key intermediate in the total synthesis of (+)-deoxoprosopinine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Cellini et al.
Microbial ecology, 79(2), 383-396 (2019-07-31)
The phyllosphere is a complex environment where microbes communicate through signalling molecules in a system, generally known as quorum sensing (QS). One of the most common QS systems in Gram-negative proteobacteria is based on the production of N-acyl homoserine lactones
Palladium-Catalyzed Intermolecular Aerobic Oxidative Amination of Terminal Alkenes: Efficient Synthesis of Linear Allylamine Derivatives
Liu Guosheng, et al.
Angewandte Chemie (International ed. in English), 47(25), 4733-4736 (2008)
Chemical composition and anti-inflammatory effects of essential oil from Farfugium japonicum flower
Kim J-Y, et al.
Journal of Oleo Science, 57(11), 623-628 (2008)
Lieuwe D J Bos et al.
PLoS pathogens, 9(5), e1003311-e1003311 (2013-05-16)
Ideally, invading bacteria are detected as early as possible in critically ill patients: the strain of morbific pathogens is identified rapidly, and antimicrobial sensitivity is known well before the start of new antimicrobial therapy. Bacteria have a distinct metabolism, part
Stereoselective strategies for the construction of polysubstituted piperidinic compounds and their applications in natural products? synthesis
Kandepedu, N, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 4(8), 1655-1704 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service