Skip to Content
Merck
All Photos(3)

Documents

241903

Sigma-Aldrich

Aluminum nitride

powder, 10 μm, ≥98%

Synonym(s):

Aluminium nitride, Azanylidynealumane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlN
CAS Number:
Molecular Weight:
40.99
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

powder

particle size

10 μm

mp

>2200 °C (lit.)

density

3.26 g/mL at 25 °C (lit.)

SMILES string

N#[Al]

InChI

1S/Al.N

InChI key

PIGFYZPCRLYGLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum Nitride(AlN) is a non-oxide semiconductor material with a hexagonal Wurtzite crystal structure. It has a wide direct bandgap of 6.12 eV. AIN is a suitable additive in making thermal grease and electric cable insulation owing to its very high thermal conductivity (300 W/(m. K)) and electrical resistivity. It is also used in microelectronics, electroluminescent devices, and sensors because of its non-toxicity, superior thermal, electrical, mechanical, and chemical properties.

Application

Aluminum Nitride can be used as a:
  • ceramic material in the preparation of nano capsulated phase change material for building thermal management systems.
  • matrix in the preparation of metacomposites along with graphene platelets to improve their thermal conductive properties.
  • foaming agent in the preparation of foam glass using glass waste.
It is also used to prepare ceramic matrix composite material along with molybdenum to generate high-energy X-rays.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel hybrid composite phase change materials with high thermal performance based on aluminium nitride and nanocapsules
Lu Wang, et al.
Energy, 238, 121775-121775 (2022)
Band structure and fundamental optical transitions in wurtzite AlN
Li, Jing, et al.
Applied Physics Letters, 83(25), 5163-5165 (2003)
Graphene platelets/aluminium nitride metacomposites with double percolation property of thermal and electrical conductivity
Yin, Rui et al.
J. Eur. Ceram. Soc., 38(14), 4701-4706 (2018)
Advanced ceramic matrix composites for high energy x-ray generation
Khan, Amir Azam and Labbe, Jean Claude
10th Internatl. Conf. on Organic Synthesis, Bangalore, India, December 1994, 2(4), 045015-045015 (2012)
Experimental investigation on thermal conductivity and viscosity of aluminum nitride nanofluid
Wei Yu,et al.
Particuology, 9(2), 187-191 (2011)

Articles

Conducting polymers such as polyaniline, polythiophene and polyfluorenes are now much in the spotlight for their applications in organic electronics and optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service