Skip to Content
Merck
All Photos(2)

Key Documents

236373

Sigma-Aldrich

Dibenzofuran

98%

Synonym(s):

Diphenylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O
CAS Number:
Molecular Weight:
168.19
Beilstein:
121100
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

154-155 °C/20 mmHg (lit.)

mp

80-82 °C (lit.)

solubility

acetic acid: soluble(lit.)
benzene: soluble(lit.)
diethyl ether: soluble(lit.)
ethanol: soluble(lit.)
water: insoluble(lit.)

SMILES string

c1ccc2c(c1)oc3ccccc23

InChI

1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

TXCDCPKCNAJMEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibenzofuran has been converted to a vacuum-sublimable, electron-transporting host material for blue-green electrophosphorescent molecule, iridium (III) bis(4,6-(di-fluorophenyl)pyridinato-N,C2′)picolinate. Biodegradation of dibenzofuran (DF) and its structural analogs by a newly isolated Agrobacterium sp. PH-08 has been reported.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T T Le et al.
Journal of applied microbiology, 116(3), 542-553 (2013-11-28)
To demonstrate the biodegradation of dibenzofuran (DF) and its structural analogs by a newly isolated Agrobacterium sp. PH-08. To assess the biodegradation potential of newly isolated Agrobacterium sp. PH-08, various substrates were evaluated as sole carbon sources in growth and
Paul A Vecchi et al.
Organic letters, 8(19), 4211-4214 (2006-09-08)
Dibenzofuran (DBF) is converted to a vacuum-sublimable, electron-transporting host material via 2,8-substitution with diphenylphosphine oxide moieties. Close pi-pi stacking and the inductive influence of P=O moieties impart favorable electron-transport properties without lowering the triplet energy. A maximum external quantum efficiency
Jannie Christensen et al.
Chemistry, an Asian journal, 8(3), 648-652 (2012-12-15)
An intramolecular, organocatalyzed Michael addition has been developed to obtain biologically important 2,3-disubstituted cis-2,3-dihydrobenzofurans. By using mandelic acid salts of primary aminocatalysts, derived from cinchona alkaloids, the intramolecular cyclization reaction has been developed to proceed in high yield, with moderate
Ruud P M Dings et al.
The Journal of pharmacology and experimental therapeutics, 344(3), 589-599 (2012-12-13)
Galectin-1 (gal-1), which binds β-galactoside groups on various cell surface receptors, is crucial to cell adhesion and migration, and is found to be elevated in several cancers. Previously, we reported on 6DBF7, a dibenzofuran (DBF)-based peptidomimetic of the gal-1 antagonist
Ulrika Nordlöf et al.
The Science of the total environment, 438, 166-173 (2012-09-25)
Concentrations of polychlorinated dibenzo-p-dioxins (PCDD), polychlorinated dibenzofurans (PCDF), and dioxin-like non-ortho-PCBs were measured in white-tailed sea eagle (WTSE) eagle eggs collected along the Swedish coast of the Baltic Sea during the period 1992-2004. The eggs represent two different subpopulations with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service