Skip to Content
Merck
All Photos(1)

Key Documents

236055

Sigma-Aldrich

Lithium aluminum hydride solution

0.5 M in 2-methoxyethyl ether

Synonym(s):

LAH, Lithium alanate, Lithium tetrahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

0.5 M in 2-methoxyethyl ether

density

0.95 g/mL at 25 °C

storage temp.

2-30°C

SMILES string

[Li+].[AlH4-]

InChI

1S/Al.Li.4H/q-1;+1;;;;

InChI key

OCZDCIYGECBNKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant or reagent for:
  • The preparation of thermoplastic polyester polyamides from oleic acid
  • Lithium-polymer batteries
  • Hydrodefluorination of gem-difluoromethylene derivatives
  • Asymmetric aldol reactions
  • Synthesis of Li-Al-N-H composites with hydrogen absorption / desorption properties

LAH is a powerful reducing agent for many different reduction reactions such as that of ketones to alcohols

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium Aluminum Hydride.
eEROS (Encyclopedia of Reagents for Organic Synthesis), 126(3), 989-992 (2001)
Solid Acids as Heterogeneous Support for Primary Amino Acid-Derived Diamines in Direct Asymmetric Aldol Reactions.
Demuynck ALW, et al.
Advanced Synthesis & Catalysis, 353(5), 725-732 (2011)
Enhanced performance in lithium-polymer batteries using surface-functionalized Si nanoparticle anodes and self-assembled block copolymer electrolytes.
Choi, Ilyoung and Ahn, Hyungmin and Park, Moon Jeong
Macromolecules, 44(18), 7327-7334 (2011)
Chen, R.;
Materials Chemistry and Physics, 126(3), 989-989 (2011)
ZnCl2-catalyzed hydrodefluorination of gem-difluoromethylene derivatives with lithium aluminum hydride.
Cheng, Jianhang and Wu, Jingjing and Cao, Song
Tetrahedron Letters, 52(27), 3481-3484 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service