Skip to Content
Merck
All Photos(2)

Key Documents

234826

Sigma-Aldrich

Bis(cyclopentadienyl)titanium(IV) dichloride

97%

Synonym(s):

Cp2TiCl2, Dichlorobis(1,3-cyclopentadiene)titanium, Dichlorodicyclopentadienyltitanium, Dichlorotitanocene, Cp2TiCl2, Di(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Cl2Ti
CAS Number:
Molecular Weight:
248.96
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: titanium
reagent type: catalyst

mp

260-280 °C (dec.) (lit.)

density

1.6 g/mL at 25 °C (lit.)

SMILES string

Cl[Ti]Cl.[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.2ClH.Ti/c2*1-2-4-5-3-1;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

XKLWATAZDMHTSH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

One-pot synthesis of α-methylene-γ-butyrolactones from benzaldehydes and bromomethylacrylates.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alberto García-Peñas et al.
Journal of inorganic biochemistry, 106(1), 100-110 (2011-11-25)
Titanocene dichloride [Ti(η(5)-C(5)H(5))(2)Cl(2)] (1), has been grafted onto dehydrated hydroxyapatite (HAP), Al(2)O(3) and two mesoporous silicas MSU-2 (Michigan State University Silica type 2) and HMS (Hexagonal Mesoporous Silica), to give the novel materials HAP/[Ti(η(5)-C(5)H(5))(2)Cl(2)] (S1) (1.01 wt.% Ti), Al(2)O(3)/[Ti(η(5)-C(5)H(5))(2)Cl(2)] (S2)
Katherine M Buettner et al.
Dalton transactions (Cambridge, England : 2003), 40(37), 9580-9588 (2011-08-19)
Since the discovery of the anticancer activity of titanocene dichloride (TDC), many derivatives have been developed and evaluated. MKT4, a soluble, water-stable formulation of TDC, was used for both Phase I and Phase II human clinical trials. This formulation is
Francesco Caruso et al.
Mini reviews in medicinal chemistry, 4(1), 49-60 (2004-02-03)
Most anticancer titanium compounds act against tumors in the gastrointestinal tract. Activity towards breast, lung and skin (melanoma) cancers is shown by some as well. Among their appealing properties is that they do not show common side effects of widely
Mauro Ravera et al.
Journal of inorganic biochemistry, 99(12), 2264-2269 (2005-10-08)
Titanocene dichloride exhibits anti-proliferative activity in a wide spectrum of murine and human tumors. Although it is still unclear as to which species are active in biological media, they all readily deliver Ti(IV) to transferrin, the protein that transports iron
Li Ming Gao et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 12(7), 959-967 (2007-06-15)
Functionalization of cyclopentadienyl (Cp) ligands and incorporation of these into a Ti(IV) center require careful design and selection of the appropriate synthetic routes to obtain the desired product in reasonably good yields. As part of our research efforts in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service