Skip to Content
Merck
All Photos(2)

Documents

232408

Sigma-Aldrich

4-Chloropyridine N-oxide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4ClNO
CAS Number:
Molecular Weight:
129.54
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

160 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[O-][n+]1ccc(Cl)cc1

InChI

1S/C5H4ClNO/c6-5-1-3-7(8)4-2-5/h1-4H

InChI key

DPJVRASYWYOFSJ-UHFFFAOYSA-N

General description

4-Chloropyridine N-oxide forms complexes with uranyl chloride and has been investigated by IR spectra, electronic spectra, molar conductivity and magnetic susceptibility measurements.

Application

4-Chloropyridine N-oxide was used as oxygen source in the preparation of [(N,N′-bis(2-methyl-2-mercaptopropane)-1,5-diazacyclooctane)Fe]2O complexes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uranyl chloride complexes of pyridine-N-oxide and 4-substituted pyridine-N-oxides: Infra-red spectra, electronic spectra and magnetic susceptibility studies.
Balakrishnan PV, et al.
J. Inorg. Nucl. Chem., 28(2), 537-541 (1966)
Ghezai Musie et al.
Inorganic chemistry, 37(16), 4086-4093 (2001-10-24)
The (&mgr;-oxo)diiron(III) complexes of N,N'-bis(2-methyl-2-mercaptopropane)-1,5-diazacyclooctane (H(2)bme-daco) and N,N'-bis(mercaptoethyl)-1,5-diazacyclooctane (H(2)bme-daco) ligands present uncommon examples of well-characterized (&mgr;-oxo)diiron(III) species in the presence of sulfur donors. Whereas the reaction of molecular oxygen with a solution of diiron(II) complex, [(bme-daco)Fe](2), affords slow formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service