Skip to Content
Merck
All Photos(3)

Documents

228656

Sigma-Aldrich

Neodymium(III) oxide

99.9% trace metals basis

Synonym(s):

Neodymia, Neodymium oxide, Neodymium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd2O3
CAS Number:
Molecular Weight:
336.48
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: neodymium

density

7.24 g/mL at 20 °C (lit.)

SMILES string

O=[Nd]O[Nd]=O

InChI

1S/2Nd.3O

InChI key

HBPPDPSYLUIKHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neodymium(III) oxide, also known as neodymia, is a rare earth metal oxide with high density. This high-purity (99.9% trace metal basis) material is widely used in ceramics as a dopant to fabricate glasses, solid-state lasers, dye-sensitized solar cells, and more. In the biomedical industry, neodymium(III) oxide is being investigated for potential applications in drug delivery systems and bioimaging due to its magnetic properties.

Application

  • Role of rare-earth oxides, conjugated with [Formula: see text], in the enhancement of power conversion efficiency of dye sensitized solar cells (DSSCs).: The paper investigates how Neodymium(III) oxide, among other rare-earth oxides, contributes to improving the power conversion efficiency in dye-sensitized solar cells, highlighting its potential in renewable energy technologies. (Sharma et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service