Skip to Content
Merck
All Photos(1)

Key Documents

224898

Sigma-Aldrich

Thallium(I) chloride

99%

Synonym(s):

Thallous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlCl
CAS Number:
Molecular Weight:
239.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder

vapor pressure

10 mmHg ( 517 °C)

Quality Level

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: thallium

mp

430 °C (lit.)

density

7 g/mL at 25 °C (lit.)

SMILES string

Cl[Tl]

InChI

1S/ClH.Tl/h1H;/q;+1/p-1

InChI key

GBECUEIQVRDUKB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare cage, dinuclear complexes (TI-cyclodiphosphazanes) which possess interesting metal-metal interactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G F Santonastaso et al.
The Science of the total environment, 630, 62-71 (2018-02-24)
Permeable Adsorptive Barriers (PABs), filled with a commercial activated carbon, are tested as a technique for the remediation of a thallium (I)-contaminated aquifer located in the south of Italy. Thallium adsorption capacity of the activated carbon is experimentally determined through
Biological safety of nasal thallium-201 administration: a preclinical study for olfacto-scintigraphy.
Kohshin Washiyama et al.
Journal of radiation research, 52(4), 450-455 (2011-09-10)
P A Howard
DICP : the annals of pharmacotherapy, 25(10), 1085-1091 (1991-10-01)
An intravenous form of dipyridamole has been approved for cardiac stress testing with thallous chloride Tl 201 (201Tl) imaging. The procedure provides an alternative method for stress testing of patients who cannot perform strenuous physical exercise because of physical limitations
Kimie Asao et al.
Annals of nuclear medicine, 26(2), 123-130 (2011-10-29)
The interpolated projection data estimation processing (IPDE) method increases the amount of projection data by interpolation of the projection data. We examined the usefulness of the IPDE method for (201)Tl myocardial perfusion imaging (MPI) single photon emission computed tomography (SPECT)
Mahbub e Khoda et al.
Journal of radiation research, 53(3), 361-367 (2012-06-29)
This study aims to investigate the efficacy of in vitro Thallium-201 Chloride (Tl-201) and in vitro and in vivo Tc-99m HYNIC-coupled Annexin V (TAV) in the early detection of radiation induced apoptosis, a proxy indicator of radiation therapy (RT) efficacy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service