Skip to Content
Merck
All Photos(3)

Documents

224677

Sigma-Aldrich

12-Bromo-1-dodecanol

99%

Synonym(s):

1-Bromo-12-dodecanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)12OH
CAS Number:
Molecular Weight:
265.23
Beilstein:
1742764
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

155 °C/4 mmHg (lit.)

mp

34-36 °C (lit.)

SMILES string

OCCCCCCCCCCCCBr

InChI

1S/C12H25BrO/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h14H,1-12H2

InChI key

ASIDMJNTHJYVQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Weis et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(46), 10946-10953 (2019-06-25)
Photoisomerization in some azobenzene-containing polymers (azopolymers) results in reversible solid-to-liquid transitions because trans- and cis-azopolymers have different glass transition temperatures. This property enables photoinduced healing and processing of azopolymers with high spatiotemporal resolution. However, a general lack of knowledge about
Yui Masuda et al.
Bioscience, biotechnology, and biochemistry, 66(7), 1531-1537 (2002-09-13)
The ceramide sex pheromone [(2S,2'R,3S,4R)-2-(2'-hydroxy-21-methyldocosanoylamino)-1,3,4-pentadecanetriol (1)] of the female hair crab (Erimacrus isenbeckii) was synthesized by starting from (S)-serine and 12-bromo-1-dodecanol.
Tetrahedron, 63, 5587-5587 (2007)
Farhana R Pinu et al.
Metabolomics : Official journal of the Metabolomic Society, 15(1), 3-3 (2019-03-05)
Although Sauvignon Blanc (SB) grapes are cultivated widely throughout New Zealand, wines from the Marlborough region are most famous for their typical varietal combination of tropical and vegetal aromas. These wines differ in composition from season to season as well
Eike T Röchow et al.
Polymers, 12(8) (2020-08-06)
Solid polymer electrolytes for bipolar lithium ion batteries requiring electrochemical stability of 4.5 V vs. Li/Li+ are presented. Thus, imidazolium-containing poly(ionic liquid) (PIL) networks were prepared by crosslinking UV-photopolymerization in an in situ approach (i.e., to allow preparation directly on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service