223328
Tungstic acid
99%
Synonym(s):
dihydroxy(dioxo)tungsten
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
H2WO4
CAS Number:
Molecular Weight:
249.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
99%
form
powder
density
5.5 g/mL at 25 °C (lit.)
SMILES string
O[W](O)(=O)=O
InChI
1S/2H2O.2O.W/h2*1H2;;;/q;;;;+2/p-2
InChI key
CMPGARWFYBADJI-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Tungstic acid may be prepared by either digesting tungstate of lime or by treating tungsten with hydrochloric acid. 1
Application
Starting material for the synthesis of a novel dimeric heteropolyoxopeoxotungstate which has potential in further studies of grafted polyoxoperoxo species on silica.
Packaging
5 g packaged in glass bottles; 100 g, 500 g packaged in poly bottles
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Piquemal, J-Y. et al.
Chemical Communications (Cambridge, England), 473-473 (1997)
J Kawada et al.
The Journal of endocrinology, 95(1), 117-124 (1982-10-01)
The role of xanthine oxidase in thyroid function was studied in the rat in vivo by different approaches. Allopurinol, an inhibitor of xanthine oxidase, was administered by mixing it with a powdered diet (16 mg/100 g body wt per day
G Soman et al.
Biochemistry, 22(21), 4994-5000 (1983-10-11)
The differential effects of the oxyanions of the early transition metals ions V(V), W(VI), and Mo(VI) on the catalytic activity and coenzyme binding of rabbit skeletal muscle phosphorylase are studied. The oligoanions of V(V), W(VI), and Mo(VI) are potent inhibitors
Bromide-assisted oxidation of substituted phenols with hydrogen peroxide to the corresponding p-quinol and p-quinol ethers over WO4(2-)-exchanged layered double hydroxides.
B F Sels et al.
Angewandte Chemie (International ed. in English), 44(2), 310-313 (2004-12-23)
S T LaBrie et al.
Molecular & general genetics : MGG, 233(1-2), 169-176 (1992-05-01)
The characterization of mutants that are resistant to the herbicide chlorate has greatly increased our understanding of the structure and function of the genes required for the assimilation of nitrate. Hundreds of chlorate-resistant mutants have been identified in plants, and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service