Skip to Content
Merck
All Photos(3)

Key Documents

219274

Sigma-Aldrich

Eicosane

99%

Synonym(s):

n-Eicosane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18CH3
CAS Number:
Molecular Weight:
282.55
Beilstein:
1700722
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

9.8 (vs air)

Assay

99%

form

solid

bp

220 °C/30 mmHg (lit.)

mp

35-37 °C (lit.)

solubility

hexane: soluble 100 mg/mL, clear

SMILES string

CCCCCCCCCCCCCCCCCCCC

InChI

1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3

InChI key

CBFCDTFDPHXCNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

n-Eicosane encapsulated in the nanosized pores of silica exhibits novel phase change behavior.

Application

Eicosane was used:
  • to fabricate a modified carbon paste electrode composed of Cu2+-mediated salbutamol-imprinted polymer nanoparticles
  • in formulation of microcapsules of n-eicosane coated with a polysiloxane

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitalii Patsula et al.
Scientific reports, 9(1), 10765-10765 (2019-07-26)
Magnetite (Fe3O4) nanoparticles with uniform sizes of 10, 20, and 31 nm were prepared by thermal decomposition of Fe(III) oleate or mandelate in a high-boiling point solvent (>320 °C). To render the particles with hydrophilic and antifouling properties, their surface was coated
Andrew J Brown et al.
Angewandte Chemie (International ed. in English), 54(20), 5864-5868 (2015-03-27)
Given the recent advent of mononuclear single-molecule magnets (SMMs), a rational approach based on lanthanides with axially elongated f-electron charge cloud (prolate) has only recently received attention. We report herein a new SMM, [Li(THF)4[Er{N(SiMe3)2}3Cl]⋅2 THF, which exhibits slow relaxation of the
Witold Fortuniak et al.
Colloid and polymer science, 291(3), 725-733 (2013-03-06)
The coemulsification method suitable for the formulation of microcapsules of
Ho Young Woo et al.
Nanomaterials (Basel, Switzerland), 11(1) (2021-01-21)
In this study, we demonstrate the colloidal synthesis of nearly monodisperse, sub-100-nm phase change material (PCM) nanobeads with an organic n-paraffin core and poly(methylmethacrylate) (PMMA) shell. PCM nanobeads are synthesized via emulsion polymerization using ammonium persulfate as an initiator and
Fernando J Guerrieri et al.
Proceedings. Biological sciences, 276(1666), 2461-2468 (2009-04-15)
Discriminating among individuals and rejecting non-group members is essential for the evolution and stability of animal societies. Ants are good models for studying recognition mechanisms, because they are typically very efficient in discriminating 'friends' (nest-mates) from 'foes' (non-nest-mates). Recognition in

Protocols

Separation of Decane; Dodecane; Tetradecane; Hexadecane; Octadecane; Eicosane; Docosane; Tetracosane; Hexacosane; Octacosane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service