Skip to Content
Merck
All Photos(2)

Key Documents

217913

Sigma-Aldrich

Tin(IV) chloride

99.995% trace metals basis

Synonym(s):

Stannic chloride fuming

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl4
CAS Number:
Molecular Weight:
260.52
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

9 (vs air)

Quality Level

vapor pressure

10 mmHg ( 10 °C)
18.6 mmHg ( 20 °C)
20 mmHg ( 22 °C)

Assay

99.995% trace metals basis

form

liquid

reaction suitability

core: tin
reagent type: Lewis acid
reagent type: catalyst

bp

114 °C (lit.)

mp

−33 °C (lit.)

density

2.226 g/mL at 25 °C (lit.)

SMILES string

Cl[Sn](Cl)(Cl)Cl

InChI

1S/4ClH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

HPGGPRDJHPYFRM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Doping polyaniline, an intensively studied polymer, with SnCl4 stabilizes its solution in nitromethane and allows solution processing.[1]
Lewis acid catalyst[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinyan Dai et al.
ChemSusChem, 12(23), 5092-5098 (2019-10-20)
One of the key challenges for the development of lithium-ion batteries is the preparation of high-performance anode materials. In this paper, a micro/nanostructured Si@SnS2 -rGO composite is reported in which Si nanoparticles with a particle size of 30 nm are electrostatically
Kulszewicz-Bajer, I. et al.
Chemistry of Materials, 11, 552-552 (1999)
Chemistry Letters (Jpn), 36, 826-826 (2007)
Bastian Waduge Naveen Harindu Hemasiri et al.
Scientific reports, 7(1), 17868-17868 (2017-12-21)
Indium tin oxide (ITO) still remains as the main candidate for high-performance optoelectronic devices, but there is a vital requirement in the development of sol-gel based synthesizing techniques with regards to green environment and higher conductivity. Graphene/ITO transparent bi-film was
Leo A Paquette et al.
The Journal of organic chemistry, 68(22), 8614-8624 (2003-10-25)
The concept of spirocyclic restriction, when generically applied to nucleoside mimics, allows for the preparation of diastereomeric pairs carrying either a syn- or anti-oriented hydroxyl at C-5'. Reported herein are convenient synthetic routes to enantiomerically pure 1-oxaspiro[4.4]nonanes featuring fully dihydroxylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service