Skip to Content
Merck
All Photos(1)

Key Documents

216879

Sigma-Aldrich

6-Aminopyridine-3-carboxylic acid

97%

Synonym(s):

6-Aminonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
Beilstein:
115992
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

>300 °C (lit.)

functional group

carboxylic acid

SMILES string

Nc1ccc(cn1)C(O)=O

InChI

1S/C6H6N2O2/c7-5-2-1-4(3-8-5)6(9)10/h1-3H,(H2,7,8)(H,9,10)

InChI key

ZCIFWRHIEBXBOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrosynthesis of 6-aminopyridine-3-carboxylic acid (6-aminonicotinic acid) by electrochemical reduction of 2-amino-5-bromo and 2-amino-5-chloropyridine in the presence of CO2 at silver electrode has been reported.

Application

6-Aminopyridine-3-carboxylic acid (6-Aminonicotinic acid) was used in the preparation of resin-bound 2-aminoazine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A three-component solid-phase synthesis of 3-aminoimidazo [1, 2-a] azines.
Blackburn C.
Tetrahedron Letters, 39(31), 5469-5472 (1998)
D J Jamieson et al.
Journal of bacteriology, 168(1), 389-397 (1986-10-01)
Expression of the tripeptide permease gene tppB is anaerobically induced. This induction is independent of the fnr (oxrA) gene product, which is known to be required for the anaerobic induction of several respiratory enzymes. We isolated, characterized, and mapped mutations
Raman Sharma et al.
Xenobiotica; the fate of foreign compounds in biological systems, 49(12), 1447-1457 (2019-02-13)
1. The absorption, metabolism, and excretion of a single oral 450-mg dose of [14C]-(S)-6-(3-cyclopentyl-2-(4-trifluoromethyl)-1H-imidazol-1-yl)propanamido)nicotinic acid (PF-04991532), a hepatoselective glucokinase activator, was investigated in humans. Mass balance was achieved with ∼94.6% of the administered dose recovered in urine and feces. The
J W Foster et al.
Journal of general microbiology, 130(11), 2873-2881 (1984-11-01)
Mutants of Salmonella typhimurium supersensitive to the nicotinic acid analogue 6-amino-nicotinic acid (6ANA) were isolated as unable to grow on what are normally subinhibitory concentrations of the analogue. The mutations were classified on the basis of their map positions as
Electrocatalytic synthesis of 6-aminonicotinic acid at silver cathodes under mild conditions.
Gennaro A, et al.
Electrochemical Communications, 6(7), 627-631 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service