Skip to Content
Merck
All Photos(1)

Documents

21240

Sigma-Aldrich

Calcium silicide

technical

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSi2
CAS Number:
Molecular Weight:
96.25
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical

Quality Level

form

powder

composition

Ca, 30-33%
Si, 60-62%

SMILES string

[Ca++].[Si-]#[Si-]

InChI

1S/Ca.Si2/c;1-2/q+2;-2

InChI key

ATJCUTDHOFKCKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao He et al.
Biomaterials, 255, 120181-120181 (2020-06-23)
Breast cancer (BC) is the most common malignant disease affecting women's health worldwide. The benefits from conventional therapeutic modalities are severely limited. An increasing number of promising photothermal materials have been recently developed and introduced into the therapeutic regimens of
Parthiban Pazhamalai et al.
ACS applied materials & interfaces, 11(1), 624-633 (2018-11-27)
Two-dimensional siloxene sheets are an emerging class of materials with an eclectic range of potential applications including electrochemical energy conversion and storage sectors. Here, we demonstrated the dehydrogenation/dehydroxylation of siloxene sheets by thermal annealing at high temperature (HT) and investigated
Xiaoliang Yan et al.
Small (Weinheim an der Bergstrasse, Germany), 16(35), e2001435-e2001435 (2020-08-06)
1D silicon-based nanomaterials, renowned for their unique chemical and physical properties, have enabled the development of numerous advanced materials and biomedical technologies. Their production often necessitates complex and expensive equipment, requires hazardous precursors and demanding experimental conditions, and involves lengthy
Karthikeyan Krishnamoorthy et al.
Nature communications, 11(1), 2351-2351 (2020-05-13)
The design and development of self-charging supercapacitor power cells are rapidly gaining interest due to their ability to convert and store energy in an integrated device. Here, we have demonstrated the fabrication of a self-charging supercapacitor using siloxene sheets as
Xiaoliang Yan et al.
Nature communications, 10(1), 2608-2608 (2019-06-15)
Two-dimensional (2D) materials are of considerable interest for catalyzing the heterogeneous conversion of CO2 to synthetic fuels. In this regard, 2D siloxene nanosheets, have escaped thorough exploration, despite being composed of earth-abundant elements. Herein we demonstrate the remarkable catalytic activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service