Skip to Content
Merck
All Photos(2)

Documents

209392

Sigma-Aldrich

3-Methoxyphenylacetonitrile

99%

Synonym(s):

3-Methoxybenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CN
CAS Number:
Molecular Weight:
147.17
Beilstein:
1865539
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

164-165 °C/20 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(CC#N)c1

InChI

1S/C9H9NO/c1-11-9-4-2-3-8(7-9)5-6-10/h2-4,7H,5H2,1H3

InChI key

LXKNAUOWEJWGTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methoxyphenylacetonitrile was used in the synthesis of:
  • new immunogen for homovanillic acid
  • β,β′-cyclobisalkylated melatoninergic phenylalkylamides
  • α-sec-butyl-3-methoxy phenylacetonitrile, antispasmodic agent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Synthesis of Simple, ?,?′-Cyclobisalkylated Melatoninergic Phenylalkylamides.
Tsotinis A, et al.
Letters in Organic Chemistry, 4(2), 92-95 (2007)
Synthesis of a homovanillic acid immunogen that incorporates an isosteric group designed to generate antibodies with improved specificity.
Gallacher G, et al.
Biogenic Amines, 11(1), 49-62 (1995)
Antispasmodic agents. 1. Syntheses and pharmacological activity of aminoalkyl 3-substituted phenylacetates.
T Kametani et al.
Journal of medicinal chemistry, 14(1), 72-75 (1971-01-01)
Suphannika Intanon et al.
Journal of agricultural and food chemistry, 62(30), 7423-7429 (2014-07-08)
Meadowfoam (Limnanthes alba Hartw. ex Benth.) is an oilseed crop grown in the Willamette Valley of Oregon. Meadowfoam seed meal (MSM), a byproduct after oil extraction, contains 2-4% glucosinolate (glucolimnanthin). Activated MSM, produced by adding freshly ground myrosinase-active meadowfoam seeds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service