Skip to Content
Merck
All Photos(2)

Key Documents

204161

Sigma-Aldrich

Ammonium perrhenate

99.999% trace metals basis

Synonym(s):

Metaperrhenic acid ammonium salt, Rhenium standard solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4ReO4
CAS Number:
Molecular Weight:
268.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
solid

Quality Level

Assay

99.999% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: rhenium

impurities

≤15.0 ppm Trace Metal Analysis

density

3.97 g/mL at 25 °C (lit.)

SMILES string

N.O[Re](=O)(=O)=O

InChI

1S/H3N.H2O.3O.Re/h1H3;1H2;;;;/q;;;;;+1/p-1

InChI key

XPSXHXWHJAIQJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reduction of this compound with 2-hydrazinopyridine and triphenylphosphine liads to rhenium(III) organohydrazides which offer potential use as radiopharmaceuticals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lingli Huang et al.
Science advances, 6(47) (2020-11-20)
Low-dimensional materials usually exhibit mechanical properties from those of their bulk counterparts. Here, we show in two-dimensional (2D) rhenium disulfide (ReS2) that the fracture processes are dominated by a variety of previously unidentified phenomena, which are not present in bulk
Perez-Lourido, P. et al.
Inorganic Chemistry, 38, 1511-1511 (1999)
Jeffrey G Linger et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(33), 12013-12018 (2014-08-06)
Lignin is an energy-dense, heterogeneous polymer comprised of phenylpropanoid monomers used by plants for structure, water transport, and defense, and it is the second most abundant biopolymer on Earth after cellulose. In production of fuels and chemicals from biomass, lignin
Yuee Feng et al.
Inorganic chemistry, 48(23), 11058-11066 (2009-11-06)
The detailed syntheses of complexes of the form [Re(O)(X)(RNCH(2)CH(2))(2)N(Me)] (X = Me, Cl, I, R = mesityl, C(6)F(5)), 1-3, incorporating diamidoamine ancillary ligands are described. X-ray crystal structures for the complexes [Re(O)(Me)((C(6)F(5))NCH(2)CH(2))(2)N(Me)], 1a, [Re(O)(I)((C(6)F(5))N CH(2)CH(2))(2)N(Me)], 3a, and [Re(O)(I)((Mes)NCH(2))(2)N(Me)], 3b, are
Julie Di Bernardo et al.
Analytical biochemistry, 415(1), 32-38 (2011-05-07)
The sodium/iodide symporter (NIS) is primarily responsible for iodide accumulation in the thyroid gland for the synthesis of thyroid hormones; however, it can also transport other lyotropic anions in the thyroid gland and nonthyroid tissues. Some NIS substrates have important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service