Skip to Content
Merck
All Photos(3)

Key Documents

203440

Sigma-Aldrich

Indium(III) chloride

99.999% trace metals basis

Synonym(s):

Indium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InCl3
CAS Number:
Molecular Weight:
221.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder or flakes (or chunks)

Quality Level

Assay

99.999% trace metals basis

form

powder or flakes (or chunks)

reaction suitability

reagent type: catalyst
core: indium

impurities

≤15.0 ppm Trace Metal Analysis

density

3.46 g/mL at 25 °C (lit.)

SMILES string

Cl[In](Cl)Cl

InChI

1S/3ClH.In/h3*1H;/q;;;+3/p-3

InChI key

PSCMQHVBLHHWTO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III) chloride is a Lewis acid catalystused in a variety of organic reactions. It also finds application in the fields of catalysis, solar cells, and optoelectronics.

Application

Indium(III) chloride can be used:
  • As a catalyst for the preparation of 1,3-dithiolanes from aldehydes and ketones.
  • For acylation of phenols, thiols, alcohols, and amines.
  • As a precursor to fabricate Indium tin oxide(ITO) thin films for dye-sensitized solar cells.
  • To prepare solution-processed oxide semiconductor thin-film transistors (OS TFTs) that can be used in display devices.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indium(III) chloride as an efficient, convenient catalyst for thioacetalization and its chemoselectivity
Sengodagounder Muthusamy, et al.
Tetrahedron Letters, 42, 359-362 (2001)
A systematic study on effects of precursors and solvents for optimization of solution-processed oxide semiconductor thin-film transistors
Keon-Hee Lim, et al.
Journal of Materials Chemistry, 5, 7768-7776 (2017)
Tetrahedron Letters, 48, 6743-6743 (2007)
Mecheril Valsan Nandakumar et al.
Chemical communications (Cambridge, England), (26), 2756-2758 (2007-06-28)
The indium-bipyridine-catalyzed, enantioselective ring-opening of meso-epoxides with aliphatic and aromatic thiols furnished 1,2-mercapto alcohols in good yields and excellent enantioselectivities; the crystal structure of the chiral catalyst reveals a pentagonal-bipyramidal coordination geometry around the indium center.
Synthesis, 1712-1712 (2007)

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service