Skip to Content
Merck
All Photos(2)

Key Documents

202096

Sigma-Aldrich

New Methylene Blue N zinc chloride double salt

Dye content 90 %

Synonym(s):

Basic blue 24, Methylene blue N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22ClN3S · 0.5ZnCl2
CAS Number:
Molecular Weight:
416.05
Colour Index Number:
52030
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 90%

mp

>240 °C (dec.) (lit.)

λmax

591 nm
630 nm (2nd)

ε (extinction coefficient)

≥16000 at 242-248 nm in H2O
≥28000 at 284-290 nm in H2O
≥36000 at 588-594 nm in H2O
≥44000 at 629-635 nm in H2O

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cl-].[Cl-].[Cl-].Cl[Zn]Cl.CCNc1cc2[s+]c3cc(NCC)c(C)cc3nc2cc1C.CCNc4cc5[s+]c6cc(NCC)c(C)cc6nc5cc4C

InChI

1S/2C18H22N3S.4ClH.Zn/c2*1-5-19-13-9-17-15(7-11(13)3)21-16-8-12(4)14(20-6-2)10-18(16)22-17;;;;;/h2*7-10,19-20H,5-6H2,1-4H3;4*1H;/q2*+1;;;;;+2/p-4

InChI key

MCBJJSJOIVJYQY-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

New Methylene Blue N zinc chloride double salt has been used as a photosensitizer for the in vitro photodynamic inactivation of plant-pathogenic fungi Colletotrichum acutatum and Colletotrichum gloeosporioides. It has also been used to determine the in vitro susceptibilities of Trichophyton mentagrophytes and Trichophyton rubrum microconidia to photodynamic antimicrobial chemotherapy (PACT).

Biochem/physiol Actions

New Methylene Blue N zinc chloride double salt also known as methylene blue N is an azo dye. It can bind, neutralize and crosslink with RNA. This causes co-precipitation of ribosomal and residual RNA with the mitochondria and ferritin masses. These events make the living, young erythrocytes as microscopically visible, dark-blue clusters and filaments, referred to as reticulum. New Methylene Blue N zinc chloride double salt helps in counting of reticulocytes (erythrocytes containing RNA).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Turgeon
Linne & Ringsrud's Clinical Laboratory Science (2011)
In Vitro Photodynamic Inactivation of Plant-Pathogenic Fungi Colletotrichum acutatum and Colletotrichum gloeosporioides with Novel Phenothiazinium Photosensitizers
de Menezes HD
Applied and Environmental Microbiology, 80, 1623-1632 (2014)
PHOTODEGRADATION OF NEW METHYLENE BLUE N IN
AQUEOUS SOLUTION USING ZINC OXIDE AND
TITANIUM DIOXIDE AS CATALYST
ALI R
Jurnal Teknologi, 45, 31-42 (2006)
Susceptibilities of the dermatophytes Trichophyton mentagrophytes and T. rubrum microconidia to photodynamic antimicrobial chemotherapy with novel phenothiazinium photosensitizers and red light
Rodrigues GB
Journal of Photochemistry and Photobiology. B, Biology, 89-94 (2012)
Gabriela Braga Rodrigues et al.
Journal of photochemistry and photobiology. B, Biology, 209, 111942-111942 (2020-07-06)
Antimicrobial photodynamic treatment (APDT) has emerged as an effective therapy against pathogenic fungi with both acquired and intrinsic resistance to commonly used antifungal agents. Success of APDT depends on the availability of effective photosensitizers capable of acting on different fungal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service