Skip to Content
Merck
All Photos(1)

Documents

190772

Sigma-Aldrich

4-Bromobenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CONH2
CAS Number:
Molecular Weight:
200.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

190-193 °C (lit.)

SMILES string

NC(=O)c1ccc(Br)cc1

InChI

1S/C7H6BrNO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H2,9,10)

InChI key

ZRWNRAJCPNLYAK-UHFFFAOYSA-N

General description

Electrochemical fluorination of 4-bromobenzamide in anhydrous HF has been investigated.

Application

4-Bromobenzamide was used in the preparation of (N-benzylpiperidin-4-yl)-4-tri-n-butylstannylbenzamide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical fluorination of aromatic compounds in anhydrous HF.
Shainyan BA and Danilevich YS.
Russ. J. Org. Chem., 42(2), 214-219 (2006)
C S John et al.
Cancer research, 55(14), 3022-3027 (1995-07-15)
The synthesis of [125I]-N-(N-benzylpiperidin-4-yl)-4-iodobenzamide (4-[125I]BP), a novel radiopharmaceutical that possesses high affinity for both sigma-1 and sigma-2 receptor subtypes, and its binding characteristics to MCF-7 breast cancer cells are described. To obtain high yields (with high specific activity) of radioiodinated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service