Skip to Content
Merck
All Photos(1)

Key Documents

189839

Sigma-Aldrich

2,3-Dihydroxybenzaldehyde

97%

Synonym(s):

1,2-Dihydroxy-3-formylbenzene, 3-Formyl-1,2-benzenediol, 3-Hydroxysalicylaldehyde, 5,6-Dihydroxybenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CHO
CAS Number:
Molecular Weight:
138.12
Beilstein:
2041781
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

104-108 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear, colorless to greenish-yellow

functional group

aldehyde

SMILES string

[H]C(=O)c1cccc(O)c1O

InChI

1S/C7H6O3/c8-4-5-2-1-3-6(9)7(5)10/h1-4,9-10H

InChI key

IXWOUPGDGMCKGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Electrochemical oxidation of 2,3-dihydroxybenzaldehyde in methanol at various pH has been studied using cyclic voltammetry and controlled-potential coulometry.

Application

2,3-Dihydroxybenzaldehyde was used in the synthesis of copper(II) complexes of Schiff bases. It was also used in the synthesis of 2-ethoxy-3-hydroxy-4-nitrobenzoic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Rakibuddin et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 15(1), 86-98 (2015-12-18)
In the present work, a graphene encapsulated ZnO/Co3O4 (GE/ZnO/Co3O4) core-shell hybrid is fabricated through a facile self-assembly approach, where the mutual electrostatic interaction force drives the ZnO/Co3O4 heteronanostructures to be fully wrapped with flexible ultrathin graphene shells. The as-prepared GE/ZnO/Co3O4
Haitao Li et al.
The Journal of biological chemistry, 280(17), 16594-16600 (2005-02-08)
Organic nitrates have been used clinically in the treatment of ischemic heart disease for more than a century. Recently, xanthine oxidase (XO) has been reported to catalyze organic nitrate reduction under anaerobic conditions, but questions remain regarding the initial precursor
H Nishioka et al.
The Journal of antibiotics, 43(12), 1586-1589 (1990-12-01)
We have screened toyocamycin as an inhibitor of phosphatidylinositol kinase. It inhibited the enzyme of A431 cell membrane with an IC50 of 3.3 micrograms/ml. Adenosine and formycin A also inhibited the enzyme, but other 6 related nucleosides did not. Although
Jong H Kim et al.
FEMS microbiology letters, 281(1), 64-72 (2008-02-21)
Activities of conventional antifungal agents, fludioxonil, strobilurin and antimycin A, which target the oxidative and osmotic stress response systems, were elevated by coapplication of certain benzo analogs (aldehydes and acids). Fungal tolerance to 2,3-dihydroxybenzaldehyde or 2,3-dihydroxybenzoic acid was found to
Hong-Chao Zhang et al.
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials, 31(10), 1503-1505 (2009-02-24)
To study the chemical constituents of Elaeocarpus sylvestris. The compounds were isolated by chromatographic methods and their structures were elucidated by physico-chemical properties and spectral analysis. Six compounds were isolated and identified as: 2-hydroxy-benzaldehyde (1), coniferyl alcohol (2), umbelliferone (3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service