All Photos(2)
About This Item
Linear Formula:
Cl2C6H3COCH3
CAS Number:
Molecular Weight:
189.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.5624 (lit.)
mp
11-13 °C (lit.)
density
1.312 g/mL at 25 °C (lit.)
functional group
chloro
ketone
SMILES string
CC(=O)c1cc(Cl)ccc1Cl
InChI
1S/C8H6Cl2O/c1-5(11)7-4-6(9)2-3-8(7)10/h2-4H,1H3
InChI key
CYNFEPKQDJHIMV-UHFFFAOYSA-N
Related Categories
General description
The standard molar enthalpy of formation of 2′,5′-dichloroacetophenone in the condensed phase was studied.
Application
2′,5′-Dichloroacetophenone was used in the synthesis of 5-chloro-2-(2-thienylthio)acetophenone.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Potential noncataleptic neuroleptic agents: Synthesis and pharmacology of 7-chloro-4-[4-(2-hydroxyethyl) piperazino]-4, 5-dihydrothieno [2, 3-b]-1-benzothiepin.
Collection of Czechoslovak Chemical Communications, 48(10), 2970-2976 (1983)
Thermochemical study of some dichloroacetophenone isomers.
Ribeiro da Silva MA and Amaral LM.
The Journal of Chemical Thermodynamics, 255-261 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service