Skip to Content
Merck
All Photos(1)

Key Documents

181374

Sigma-Aldrich

Polybutadiene, cis

average Mw 200,000-300,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH=CHCH2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

98% cis

Quality Level

form

slab/chunk

mol wt

average Mw 200,000-300,000

viscosity

40, Mooney(lit.)

transition temp

Tg −102 °C

density

0.9 g/mL at 25 °C (lit.)

SMILES string

C\C=C\C

InChI

1S/C4H8/c1-3-4-2/h3-4H,1-2H3/b4-3+

InChI key

IAQRGUVFOMOMEM-ONEGZZNKSA-N

General description

Polybutadiene, cis (PB-cis) is a synthetic rubber that is prepared by solution polymerization of 1,4-butadiene. Its mechanical properties include low temperature flexibility and abrasion resistance. It can be cured by using sulfur based donors and peroxides and thus PB-cis acts as a highly cross-linked polymer with high resilience.

Application

PB-cis can be polymerized with styrene for the formation of high impact polystyrene (HIPS). It may also be used as a toughening agent for acrylonitrile butadiene rubber (ABS) based resins.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Poylmer-Liquid Interaction Parameters and Solubility Parameters (2018)
Rubber Basics, 80-81 (2002)
Morphological, mechanical properties, and fracture behavior of bulk-made ABS resins toughened by high-cis polybutadiene rubber
Yu Z, et al.
Polym. Eng. Sci., 50(5), 961-969 (2010)
Role of the rubber particle and polybutadiene cis content on the toughness of high impact polystyrene
Rovere J, et al.
Journal of materials science & technology, 43(3), 952-959 (2008)
Dong Wu et al.
Journal of the American Chemical Society, 132(45), 15936-15938 (2010-10-28)
Single-molecule force spectroscopy is used to observe the irreversible extension of a gem-dibromocyclopropane (gDBC)-functionalized polybutadiene under tension, a process akin to polymer necking at a single-molecule level. The extension of close to 28% in the contour length of the polymer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service