181285
Poly(acrylic acid)
average Mv ~450,000
Synonym(s):
PAA
About This Item
Recommended Products
crosslinking
~0.1 % cross-linked
mol wt
average Mv ~450,000
viscosity
350-2500 cP
transition temp
Tg 106 °C
SMILES string
[Na+].[O-]C(=O)C=C
InChI
1S/C3H4O2.Na/c1-2-3(4)5;/h2H,1H2,(H,4,5);/q;+1/p-1
InChI key
NNMHYFLPFNGQFZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Other applications may include:
- to study solute diffusion in Polyvinyl alcohol/PAA copolymer hydrogel
- synthesizing poly(N-isopropylacrylamide)-block-PAA copolymer which responds to both temperature and pH stimuli
- in preparing block copolymer of oligo (methyl methacrylate)/PAA for micellar delivery of hydrophobic drugs
- as thickening agent for adhesives.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.
Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.
Laboratory safety tools: Offers educational tools for chemistry, prioritizing customer health and safety, with online support available.
We have discussed shortly the applications of 1:1 and 2:1 phyllosilicate material in membrane technology, as well as the basic geometric understanding of their layered structures.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service