Skip to Content
Merck
All Photos(3)

Key Documents

178055

Sigma-Aldrich

N-Phenyl-2-naphthylamine

97%

Synonym(s):

N-(2-Naphthyl)aniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHC6H5
CAS Number:
Molecular Weight:
219.28
Beilstein:
2211188
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

395-395.5 °C (lit.)

mp

105-108 °C (lit.)

λmax

219 nm

SMILES string

N(c1ccccc1)c2ccc3ccccc3c2

InChI

1S/C16H13N/c1-2-8-15(9-3-1)17-16-11-10-13-6-4-5-7-14(13)12-16/h1-12,17H

InChI key

KEQFTVQCIQJIQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

[Analysis of nitrogen containing aging-preventive agents in vulcanizing vapors].
E Schmidt et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 29(8), 449-452 (1983-08-01)
Low sensitization rate of naphthyl mix.
W G van Ketel
Contact dermatitis, 9(1), 77-77 (1983-01-01)
Tianxiang Li et al.
Materials (Basel, Switzerland), 11(5) (2018-05-23)
With the aim of improving the anti-aging properties of nitrile-butadiene rubber (NBR), a functional organic filler, namely LDH⁻SAS, prepared by intercalating 4-amino-benzenesulfonic acid monosodium salt (SAS) into layered double hydroxides (LDHs) through anion exchange, was added to nitrile-butadiene rubber (NBR)
[Industrial toxicologic evaluation of vulcanizing vapors with special reference to nitrogen-containing aging-preventive agents].
E Schmidt et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 29(8), 452-456 (1983-08-01)
Unusual allergic contact dermatitis to aromatic amines.
L Condé-Salazar et al.
Contact dermatitis, 17(1), 42-44 (1987-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service