Skip to Content
Merck

Skip To

175757

1-Butyl alcohol-OD

98 atom % D

Synonym(s):

1-Butan(ol-d), 1-Butanol-OD, Butyl alcohol-d

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3(CH2)3OD
CAS Number:
Molecular Weight:
75.13
UNSPSC Code:
12352202
PubChem Substance ID:
EC Number:
225-201-2
MDL number:
Isotopic purity:
98 atom % D
Assay:
99% (CP)
Mass shift:
M+1
Form:
liquid
Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service


isotopic purity

98 atom % D

Quality Segment

assay

99% (CP)

form

liquid

refractive index

n20/D 1.3975 (lit.)

bp

118 °C (lit.)

density

0.821 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OCCCC

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i5D

InChI key

LRHPLDYGYMQRHN-UICOGKGYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact
Stable Isotopes Customer Service.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
175765615080615110
description

98 atom % D

description

99 atom % D

description

98 atom % D

description

≥98 atom % D, ≥99% (CP)

assay

99% (CP)

assay

99% (CP)

assay

99% (CP)

assay

≥99% (CP)

mass shift

M+1

mass shift

M+1

mass shift

M+1

mass shift

M+1

isotopic purity

98 atom % D

isotopic purity

99 atom % D

isotopic purity

98 atom % D

isotopic purity

≥98 atom % D

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

-

form

liquid

form

liquid

form

-

form

-

refractive index

n20/D 1.3975 (lit.)

refractive index

n20/D 1.3847 (lit.)

refractive index

n20/D 1.3752 (lit.)

refractive index

-


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters