Skip to Content
Merck
All Photos(2)

Key Documents

173738

Sigma-Aldrich

3,3′-Diethylthiacarbocyanine iodide

Dye content 95 %

Synonym(s):

3-Ethyl-2-(3-[3-ethyl-2(3H)benzothiazolylidene]-1-propenyl)benzothiazolium iodide, DiSC2(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21IN2S2
CAS Number:
Molecular Weight:
492.44
Beilstein:
4117818
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, 95%

mp

268-270 °C (dec.) (lit.)

λmax

560 nm

SMILES string

[I-].CCN1\C(Sc2ccccc12)=C\C=C\c3sc4ccccc4[n+]3CC

InChI

1S/C21H21N2S2.HI/c1-3-22-16-10-5-7-12-18(16)24-20(22)14-9-15-21-23(4-2)17-11-6-8-13-19(17)25-21;/h5-15H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

VZBILKJHDPEENF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

3,3′-Diethylthiacarbocyanine iodide (DTCI) is a cyanine dye, which acts as a donor in the electrochemical system. It can be used in the development of electrochemical cells.

Application

DTCI may be incorporated as a layer on gold nanorods, which can be used in surface enhanced resonance Raman spectroscopy (SERRS). It may be coated on a fused silica wafer, which can potentially be used in the development of solid state dye doped polymeric laser.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorescent sensitized fluorescent solid-state near-infrared light-emitting electrochemical cells.
Ho C, et al.
Physical Chemistry Chemical Physics, 13(39), 17729-17736 (2011)
Reconfigurable solid-state dye-doped polymer ring resonator lasers.
Chandrahalim H and Fan X
Scientific reports, 5(13), 18310-18310 (2015)
Alexander J Thompson et al.
The journal of physical chemistry. B, 119(32), 10170-10179 (2015-07-21)
Changes in microscopic viscosity represent an important characteristic of structural transitions in soft matter systems. Here we demonstrate the use of molecular rotors to explore the changes in microrheology accompanying the transition of proteins from their soluble states into a
Controlled side-by-side assembly of gold nanorods and dye molecules into polymer-wrapped SERRS-active clusters.
McLintock A, et al.
Chemical Communications (Cambridge, England), 47(13), 3757-3759 (2011)
L Marcus Wilhelmsson et al.
Nucleic acids research, 30(2), E3-E3 (2002-01-15)
As the relationship between human genes and various malfunctions and diseases becomes revealed at an ever-increasing pace, the need arises for the development of rapid genetic screening methods for diagnostic purposes. Genetic diseases show great diversity. Some are caused by

Articles

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

Developed in the last several years, fluorescence quenching microscopy (FQM) has enabled rapid, inexpensive, and high-fidelity visualization of two-dimensional (2D) materials such as graphene-based sheets and MoS2.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service