Skip to Content
Merck
All Photos(1)

Documents

17249

Sigma-Aldrich

α-Methyl-L-proline

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
4350211
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

C[C@]1(CCCN1)C(O)=O

InChI

1S/C6H11NO2/c1-6(5(8)9)3-2-4-7-6/h7H,2-4H2,1H3,(H,8,9)/t6-/m0/s1

InChI key

LWHHAVWYGIBIEU-LURJTMIESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Thaisrivongs et al.
Journal of medicinal chemistry, 30(3), 536-541 (1987-03-01)
A structure-activity analysis of peptides containing backbone C alpha-methyl modification at the P4 site of the angiotensinogen sequence led to the discovery of potent renin inhibitors with apparent in vitro metabolic stability. Boc-alpha-MePro-Phe-His-Leu psi[CHOHCH2]Val-Ile-Amp dicitrate (Va) is a potent inhibitor
Alessandro Moretto et al.
Biopolymers, 89(5), 465-470 (2007-09-07)
Methylation at the C(alpha)-position of a Pro residue was expected to lock the preceding tertiary amide (omega) torsion angle of the resulting (alphaMe)Pro to the trans disposition and to restrict the phi,psi surface to the single region where the 3(10)/alpha-helices
G Lubec et al.
Life sciences, 57(24), 2245-2252 (1995-01-01)
Collagen type I is the major protein of bone matrix and significantly reduced in osteoporosis. We tested the effect of alpha - methyl - proline on collagen synthesis in the model of the ovariectomized rat. Collagen synthesis was studied at
J H Welsh et al.
FEBS letters, 297(3), 216-220 (1992-02-10)
The ability of (S)-alpha-methylproline (alpha-MePro) to stabilise reverse-turn conformations in the peptide hormone bradykinin (BK = Arg1-Pro2-Pro3-Gly4-Phe5-Ser6-Pro7-Phe8-Arg9) has been investigated. Two BK analogues containing alpha-MePro at position 3 or position 7 were synthesised and their conformations in aqueous solution investigated
Guillem Revilla-López et al.
Biopolymers, 98(2), 98-110 (2011-09-08)
The structural consequences derived from the incorporation of either a methyl or a phenyl group at the α carbon of proline were recently investigated by quantum mechanical calculations (J Org Chem 2008, 73, 3418). In this work, the effect produced

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service