All Photos(2)
About This Item
Linear Formula:
NCC(NH2)=C(NH2)CN
CAS Number:
Molecular Weight:
108.10
Beilstein:
878333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
powder
mp
178-179 °C (lit.)
functional group
amine
nitrile
SMILES string
N\C(C#N)=C(/N)C#N
InChI
1S/C4H4N4/c5-1-3(7)4(8)2-6/h7-8H2/b4-3-
InChI key
DPZSNGJNFHWQDC-ARJAWSKDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Supplementary Hazards
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marta Ruiz-Bermejo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(49), 11437-11455 (2019-08-03)
HCN polymers are a group of complex and heterogeneous substances that are widely known in the fields of astrobiology and prebiotic chemistry. In addition, they have recently received considerable attention as potential functional material coatings. However, the real nature and
Huan Feng et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(8), 1800397-1800397 (2018-08-22)
Diagnosis and early assessment of the treatment response of rheumatoid arthritis (RA) necessitates a reliable bioanalytical method for rapid, sensitive, and specific detection of the hypochlorous acid (HOCl) biomarker in inflammatory diseases. Herein, two fluorescence probes, Probe-1 and Probe-2 are
Highly sensitive two-photon chromophores applied to three-dimensional lithographic microfabrication: design, synthesis and characterization towards two-photon absorption cross section.
Lu Y, et al.
Journal of Materials Chemistry, 14(1), 75-80 (2004)
Rafał Szabla et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(9), 2515-2521 (2014-01-29)
The photoinduced isomerization of diaminomaleonitrile (DAMN) to diaminofumaronitrile (DAFN) was suggested to play a key role in the prebiotically plausible formation of purine nucleobases and nucleotides. In this work we analyze two competitive photoisomerization mechanisms on the basis of state-of-the-art
Tong Zhao et al.
Carbohydrate polymers, 218, 163-169 (2019-06-22)
In this study, the effects of natural fermentation on the multi-scale structure and physicochemical properties of wheat starch were investigated. The results showed that fermentation reduced weight-average molecular weight (Mw) and slightly eroded the starch granule surface. An obvious increase
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service