Skip to Content
Merck
All Photos(1)

Key Documents

159441

Sigma-Aldrich

Diethyl acetylenedicarboxylate

95%

Synonym(s):

Diethyl 2-butynedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOC≡CCOOC2H5
CAS Number:
Molecular Weight:
170.16
Beilstein:
743166
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.443 (lit.)

bp

107-110 °C/11 mmHg (lit.)

density

1.063 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CCOC(=O)C#CC(=O)OCC

InChI

1S/C8H10O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-4H2,1-2H3

InChI key

STRNXFOUBFLVIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl acetylenedicarboxylate is a protein cross-linker.

Diethyl acetylenedicarboxylate is used as a Michael acceptor for O-vinyl oximes synthesis, and is used in the nucleophilic addition reaction.

Application

Diethyl acetylenedicarboxylate was used in the synthesis of:
  • 3,4,5-trisubstituted 2(5H)-furanone derivatives
  • highly functionalized thiazolidinone derivatives
  • novel cyclic peroxide glucosides
  • 4,11-dimesitylbisanthene, soluble bisanthene derivative, via Diels-Alder reaction

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdelmadjid Benmohammed et al.
Molecules (Basel, Switzerland), 19(3), 3068-3083 (2014-03-13)
We present herein the synthesis in good yields of two series of highly functionalized thiazolidinone derivatives from the reactions of various 4-phenyl-3-thio-semicarbazones with ethyl 2-bromoacetate and diethyl acetylenedicarboxylate, respectively.
Eric H Fort et al.
Journal of the American Chemical Society, 131(44), 16006-16007 (2009-10-17)
A soluble bisanthene derivative, 4,11-dimesitylbisanthene, has been synthesized in three steps from bianthrone. In hot toluene, this bisanthene undergoes a clean Diels-Alder reaction with diethyl acetylenedicarboxylate to give a rearomatized 1:1 cycloadduct and, more slowly, a rearomatized 2:1 cycloadduct. In
Microwave Induced Stereoselective Synthesis of O--Vinyl Oximes using Acetylenic Esters as Efficient Michael Acceptors
Ankush M et al.
ChemistrySelect, 3, 9464-9468 (2018)
Di-Zao Li et al.
Journal of Asian natural products research, 11(7), 613-620 (2010-02-26)
Four novel cyclic peroxide glucosides 15a, 15b, 16a, and 16b, optically pure analogs of shuangkangsu (1), which is an anti-virus natural product with an unusual skeleton isolated from the buds of Lonicera japonica Thunb, were first synthesized totally in six
Synthesis of pyrrolidinone derivatives from aniline, an aldehyde and diethyl acetylenedicarboxylate in an ethanolic citric acid solution under ultrasound irradiation
Hamideh A et al.
Green Chemistry, 18, 3582-3593 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service