Skip to Content
Merck
All Photos(1)

Documents

159425

Sigma-Aldrich

9-Chloroacridine

97%

Synonym(s):

9-Chloroacridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8ClN
CAS Number:
Molecular Weight:
213.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

116-120 °C (lit.)

SMILES string

Clc1c2ccccc2nc3ccccc13

InChI

1S/C13H8ClN/c14-13-9-5-1-3-7-11(9)15-12-8-4-2-6-10(12)13/h1-8H

InChI key

BPXINCHFOLVVSG-UHFFFAOYSA-N

Application

9-Chloroacridine was employed as chromogenic reagent in the spectrophotometric method for the quantitative determination of dapsone. It was also used in the synthesis of:
  • series of novel chalcones bearing acridine moiety attached to the amino group in their ring A
  • new acridine derivatives
  • 9-phenoxyacridine and 4-phenoxyfuro[2,3-b]quinoline derivatives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

V Tomar et al.
European journal of medicinal chemistry, 45(2), 745-751 (2009-12-22)
A series of novel chalcones bearing acridine moiety attached to the amino group in their ring A have been synthesized through noncatalyzed nucleophilic aromatic substitution reaction between various 3'-aminochalcone or 4'-aminochalcones and 9-chloroacridine. The synthesized chalcone derivatives have been characterized
Yeh-Long Chen et al.
Bioorganic & medicinal chemistry, 11(18), 3921-3927 (2003-08-21)
Mast cells, neutrophils and macrophages are important inflammatory cells that have been implicated in the pathogenesis of acute and chronic inflammatory diseases. To explore a novel anti-inflammatory agent, we have synthesized certain 9-phenoxyacridine and 4-phenoxyfuro[2,3-b]quinoline derivatives and evaluated their anti-inflammatory
Jelena Rupar et al.
Bioelectrochemistry (Amsterdam, Netherlands), 135, 107579-107579 (2020-06-14)
The electrochemical behavior of 9-chloroacridine (9Cl-A), a precursor molecule for synthesis of acridine derivatives with cytostatic activity, is a complex, pH-dependent, diffusion-controlled irreversible process. Oxidation of 9Cl-A initiates with the formation of a cation radical monomer, continues via the formation
Paula Bosch et al.
Materials (Basel, Switzerland), 12(18) (2019-09-22)
A second-generation poly(propylene imine) dendrimer modified with acridine and its Cu(II) complex have been synthesized for the first time. It has been found that two copper ions form complexes with the nitrogen atoms of the dendrimeric core by coordinate bonds.
Arumugasamy Elangovan et al.
Organic & biomolecular chemistry, 2(21), 3113-3118 (2004-10-27)
Electrogenerated chemiluminescence (ECL) of six new ethyne-based acridine derivatives (1-6) has been studied. The new acridine derivatives were synthesized by cross-coupling of 9-chloroacridine and corresponding donor-substituted phenylethynes under modified Sonogashira conditions. The donor groups were varied in the order of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service