Skip to Content
Merck
All Photos(3)

Key Documents

157716

Sigma-Aldrich

2,6-Diisopropylaniline

technical grade, 90%

Synonym(s):

2,6-Bis(1-methylethyl)benzenamine, 2,6-Bis(propan-2-yl)aniline, 2,6-Diisopropylphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2C6H3NH2
CAS Number:
Molecular Weight:
177.29
Beilstein:
2208763
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

90%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

257 °C (lit.)

mp

−45 °C (lit.)

density

0.94 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cccc(C(C)C)c1N

InChI

1S/C12H19N/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9H,13H2,1-4H3

InChI key

WKBALTUBRZPIPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Diisopropylaniline on condensation with triacetylmethane in toluene in the presence of p-toluenesulfonic acid provides 3-[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione.

Application

2,6-Diisopropylaniline was used in the preparation of multitopic Schiff-base ligand precursors. It was also used in the preparation of NSN-donor proligand, 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remote activation of nickel catalysts for ethylene oligomerization.
Yaofeng Chen et al.
Angewandte Chemie (International ed. in English), 44(7), 1108-1112 (2005-01-13)
Balamurugan Vidjayacoumar et al.
Dalton transactions (Cambridge, England : 2003), 41(26), 8175-8189 (2012-05-09)
A rigid NSN-donor proligand, 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene (H(2)[TXA(2)], 1) was prepared by palladium-catalyzed coupling of 2,6-diisopropylaniline with 4,5-dibromo-2,7-di-tert-butyl-9,9-dimethylthioxanthene. Deprotonation of 1 using (n)BuLi provided Li(2)(DME)(2)[TXA(2)] (2), and subsequent reaction with UCl(4) afforded [Li(DME)(3)][(TXA(2))UCl(3)] (4). The analogous NON-donor ligated complex [(XA(2))UCl(3)K(DME)(3)] [3; XA(2)
Yilin Sun et al.
Dalton transactions (Cambridge, England : 2003), 41(32), 9682-9688 (2012-07-12)
Four multitopic Schiff-base ligand precursors were synthesized via condensation of 4,4'-diol-3,3'-diformyl-1,1'-diphenyl or 1,3,5-tris(4-hydroxy-5-formylphenyl)benzene with 2,6-diisopropylaniline or 2,6-dimethylaniline. Amine elimination reactions of Ln[N(SiMe(3))(2)](3) (Ln = La, Nd, Sm or Y) with these multitopic ligand precursors gave ten heterogeneous rare-earth metal catalysts.
D A Razborov et al.
Dalton transactions (Cambridge, England : 2003), 44(47), 20532-20541 (2015-11-10)
In the presence of formic acid, acenaphthenequinone (AQ) reacts with one molar equivalent of 2,6-diisopropylaniline in toluene to give monoiminoacenaphtheneone (3, dpp-mian) in good yield. Reduction of compound 3 with an excess of magnesium in thf results in green crystalline
Luka A Wright et al.
Dalton transactions (Cambridge, England : 2003), 44(16), 7230-7241 (2015-03-20)
The 2-(2′-aniline)-6-imine-pyridines, 2-(C6H4-2′-NH2)-6-(CMe=NAr)C5H3N (Ar = 4-i-PrC6H4 (HL1a), 2,6-i-Pr2C6H3 (HL1b)), have been synthesised via sequential Stille cross-coupling, deprotection and condensation steps from 6-tributylstannyl-2-(2-methyl-1,3-dioxolan-2-yl)pyridine and 2-bromonitrobenzene. The palladium(II) acetate N,N,N-pincer complexes, [{2-(C6H4-2′-NH)-6-(CMe=NAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (1a), 2,6-i-Pr2C6H3 (1b)), can be prepared by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service