156655
trans-1-Decalone
98%
Synonym(s):
trans-Decahydro-1-naphthalenone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
98%
bp
73 °C/1 mmHg (lit.)
mp
30-32 °C (lit.)
SMILES string
[H][C@]12CCCC[C@]1([H])C(=O)CCC2
InChI
1S/C10H16O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h8-9H,1-7H2/t8-,9+/m1/s1
InChI key
AFEFRXAPJRCTOW-BDAKNGLRSA-N
Application
The product has been used to study its proton resonance spectra using NMR.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
195.8 °F - closed cup
Flash Point(C)
91 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Proton chemical shifts in NMR. Part 13. Proton chemical shifts in ketones and the magnetic anisotropy and electric field effect of the carbonyl group.
Journal of the Chemical Society, 2(3), 441-448 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service