Skip to Content
Merck
All Photos(3)

Key Documents

155209

Sigma-Aldrich

Pyrrolidone hydrotribromide

97%

Synonym(s):

PHT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H7NO)3 · HBr3
CAS Number:
Molecular Weight:
496.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
97%

Quality Level

Assay

97%

form

solid

mp

88-90 °C (lit.)

storage temp.

2-8°C

SMILES string

Br[BrH]Br.O=C1CCCN1.O=C2CCCN2.O=C3CCCN3

InChI

1S/3C4H7NO.Br3H/c3*6-4-2-1-3-5-4;1-3-2/h3*1-3H2,(H,5,6);3H

InChI key

ITDWTSPGISAGCS-UHFFFAOYSA-N

General description

Pyrrolidone hydrotribromide has high selectivity for ketones.

Application

Pyrrolidone hydrotribromide was used as a brominating agent.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theresa Bucsics et al.
PloS one, 13(1), e0189414-e0189414 (2018-01-10)
Transjugular intrahepatic portosystemic shunts (TIPS) are used in patients with cirrhosis for the prevention of variceal rebleeding. We retrospectively evaluated re-bleeding rate, patency, mortality, and transplant-free survival (TFS) in cirrhotic patients receiving TIPS implantation for variceal bleeding between 1994-2014. 286
Ryan J Spencer et al.
Journal of the American Society of Echocardiography : official publication of the American Society of Echocardiography, 30(10), 966-973 (2017-08-22)
Detecting bioprosthetic mitral valve dysfunction on transthoracic echocardiography can be challenging because of acoustic shadowing of regurgitant jets and a wide normal range of transvalvular gradients. Several studies in mechanical mitral valves have demonstrated the utility of the transthoracically derived
Bjoern Zante et al.
PloS one, 13(10), e0205309-e0205309 (2018-10-06)
Cardiac surgery with the use of cardiopulmonary bypass is known to induce distinct metabolic changes. Respective changes in acid-base status including increased systemic lactate levels were previously related to clinical outcomes, but data remain controversial. Therefore, we aim to investigate
Pyrrolidone hydrotribromide: a brominating agent with selectivity for ketones.
Canadian Journal of Chemistry, 47(4), 706-709 (1969)
Lor Huai Chong et al.
ACS omega, 4(9), 13902-13912 (2019-09-10)
The clinical use of some drugs, such as carbamazepine, phenytoin, and allopurinol, is often associated with adverse cutaneous reactions. The bioactivation of drugs into immunologically reactive metabolites by the liver is postulated to be the first step in initiating a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service