Skip to Content
Merck
All Photos(1)

Key Documents

15140

Sigma-Aldrich

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride

≥97.0% (AT)

Synonym(s):

BOP-Cl, Phosphoric acid bis(2-oxooxazolidide) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8ClN2O5P
CAS Number:
Molecular Weight:
254.56
Beilstein:
3654596
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (AT)

form

solid

mp

191 °C (dec.) (lit.)

SMILES string

ClP(=O)(N1CCOC1=O)N2CCOC2=O

InChI

1S/C6H8ClN2O5P/c7-15(12,8-1-3-13-5(8)10)9-2-4-14-6(9)11/h1-4H2

InChI key

KLDLRDSRCMJKGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide.

Other Notes

Reagent for activating the carboxylic group, synthesis of amides ; Esters ; Peptides

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Cabre et al.
Synthesis, 413-413 (1984)
S. Danishefsky et al
Tetrahedron Letters, 25, 4203-4203 (1984)
Zhao Yanbin et al.
Biochemical and biophysical research communications, 516(1), 102-111 (2019-06-16)
Here, we firstly examined the proliferation and invasion capacities of different osteosarcoma (OS) cell lines, and analyzed the profiling of the differentially expressed circular RNAs(circRNAs). Then, we identified a novel circRNA(circ_101356/circ_0004846, we named circSAMD4 in the present study) and found
Yanjie Xu et al.
Molecules (Basel, Switzerland), 10(1), 259-264 (2007-11-17)
Hexadepsipeptide 2, the precursor of Hirsutellide A (1), was synthesized in an overall yield of 45% from N-Boc-Me-Gly via three coupling reactions using dicyclohexylcarbodiimide (DCC), O-(7-azabenzotriazol-1-yl)-N,N,N',N'-tetramethyl- uronium hexafluorophosphate (HATU) and bis(2-oxo-3-oxazolidinyl)phosphinic chloride (BOP-Cl), respectively.
R.D. Tung et al.
Journal of the American Chemical Society, 107, 4342-4342 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service