Skip to Content
Merck
All Photos(1)

Key Documents

148970

Sigma-Aldrich

Ethyl decanoate

ReagentPlus®, ≥99%

Synonym(s):

Capric acid ethyl ester, Ethyl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOC2H5
CAS Number:
Molecular Weight:
200.32
Beilstein:
1762128
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.9 (vs air)

product line

ReagentPlus®

Assay

≥99%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

245 °C (lit.)

solubility

H2O: insoluble
alcohol: miscible
chloroform: miscible
diethyl ether: miscible

density

0.862 g/mL at 25 °C

SMILES string

CCCCCCCCCC(=O)OCC

InChI

1S/C12H24O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h3-11H2,1-2H3

InChI key

RGXWDWUGBIJHDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl decanoate has ethereal, fruity, aromatic note that is considered to be essential for a fine wine flavor.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Rodríguez-Lerma et al.
Journal of food science, 76(6), M346-M352 (2012-03-16)
A procedure for designing starter cultures for fermentation is illustrated for prickly pear wine production. The illustration includes kinetic studies on inoculated and spontaneous fermentation, microorganism identification studies based on molecular biology tools, and microbial ecology studies, which led to
Lei Bian et al.
Journal of economic entomology, 111(2), 629-636 (2018-01-24)
The tea leafhopper, Empoasca onukii Matsuda, is a serious pest of the tea plant. E. onukii prefers to inhabit vigorously growing tender tea leaves. The host selection of E. onukii adults may be associated with plant volatile compounds (VOCs). We
Yan-Qing Fu et al.
Food chemistry, 331, 127258-127258 (2020-06-17)
Flavor stability is important for the quality of tea beverages. Baking is a typical processing technology to improve the flavor of tea leaves. In present study, seven raw tea materials, including steamed spring and autumn tea leaves, pan-fired spring tea
Sumin Lee et al.
Colloids and surfaces. B, Biointerfaces, 47(1), 78-84 (2006-01-13)
The relationship between the kinetics of the lipase-catalyzed oil hydrolysis and the surface area distribution of oil droplets was investigated using ethyl decanoate and gum Arabic (GA) as a model oil and an emulsifier, respectively. Along an ethyl decanoate concentration
F B Whitfield et al.
The Journal of dairy research, 67(4), 561-569 (2000-01-11)
Analysis by gas chromatography-mass spectrometry of pasteurized milk with a fruity (pineapple like) off odour and a sour, rancid and soapy taste indicated the presence of concentrations at microg/ml levels of ethyl butanoate, ethyl hexanoate, ethyl octanoate, ethyl decanoate, octanoic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service