Skip to Content
Merck
All Photos(6)

Key Documents

147532

Sigma-Aldrich

1,3,5-Benzenetricarbonyl trichloride

98%

Synonym(s):

Benzene-1,3,5-tricarbonyl chloride, Trimesic acid trichloride, Trimesoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(COCl)3
CAS Number:
Molecular Weight:
265.48
Beilstein:
2940936
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

bp

180 °C/16 mmHg (lit.)

mp

32-38 °C (lit.)

solubility

chloroform: soluble 10%, clear to very slightly hazy, colorless to faintly yellow

density

1.487 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1cc(cc(c1)C(Cl)=O)C(Cl)=O

InChI

1S/C9H3Cl3O3/c10-7(13)4-1-5(8(11)14)3-6(2-4)9(12)15/h1-3H

InChI key

UWCPYKQBIPYOLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3,5-Benzenetricarbonyl trichloride reacts with propargyl alcohol to yield triprop-2-ynyl benzene-1,3,5-tricarboxylate.

Application

1,3,5-Benzenetricarbonyl trichloride was used in the synthesis of chiral azoaromatic dendrimeric systems. It was used to study the structure of activated composite membranes containing organophosphorus extractants as carriers. It was the starting material for two tritopic amides derived from 3- and 4-methylaminopyridine which self-assembled into nanoballs on treatment with palladium(II) nitrate in DMSO.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sushobhan Ghosh et al.
The Journal of organic chemistry, 71(22), 8412-8416 (2006-10-27)
The design and self-assembly of two new flexible supramolecular nanoballs are described. These assemblies incorporate two flexible tritopic amide and ester building blocks and were prepared in excellent yields (96-97%) via coordination driven self-assembly. The first resulted from the reaction
Luigi Angiolini et al.
Chirality, 22(1), 99-109 (2009-04-24)
New chiral azoaromatic dendrimeric systems have been synthesized starting from 1,3,5-benzenetricarbonyl trichloride as the core molecule. The simultaneous presence of the (S)-3-hydroxy pyrrolidinyl ring as the optically active moiety and the azobenzene donor-acceptor conjugated system as the photochromic group with
Zhe Yang et al.
Environmental science & technology, 52(16), 9341-9349 (2018-07-26)
Conventional thin-film composite (TFC) membranes suffer from the trade-off relationship between permeability and selectivity, known as the "upper bound". In this work, we report a high performance thin-film composite membrane prepared on a tannic acid (TA)-Fe nanoscaffold (TFCn) to overcome
Behnam Khorshidi et al.
Scientific reports, 8(1), 784-784 (2018-01-18)
The development of nano-enabled composite materials has led to a paradigm shift in the manufacture of high-performance nanocomposite membranes with enhanced permeation, thermo-mechanical, and antibacterial properties. The major challenges to the successful incorporation of nanoparticles (NPs) to polymer films are
Machodi Mathaba et al.
Membranes, 10(3) (2020-03-28)
Acid mine drainage is an environmental problem associated with mining operations and activities. Its treatment is essential to achieving environmental sustainability. In this study, a polyethersulphone (PES) membrane infused with chitosan is proposed as a point-of-use material for treating acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service